Skip to Content
Merck
All Photos(3)

Key Documents

537683

Sigma-Aldrich

2-Heptanone

99%

Synonym(s):

Methyl pentyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COCH3
CAS Number:
Molecular Weight:
114.19
Beilstein:
1699063
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
technique(s):
gas chromatography (GC): suitable
bp:
149-150 °C (lit.)
vapor pressure:
2.14 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3.94 (vs air)

Quality Level

vapor pressure

2.14 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

739 °F

expl. lim.

1.11 %, 65 °F
7.9 %, 121 °F

technique(s)

gas chromatography (GC): suitable

impurities

≤0.05% (water)

refractive index

n20/D 1.408 (lit.)

bp

149-150 °C (lit.)

mp

−35 °C (lit.)

solubility

water: soluble 4.21 g/L at 20 °C

density

0.82 g/mL at 25 °C (lit.)

SMILES string

CCCCCC(C)=O

InChI

1S/C7H14O/c1-3-4-5-6-7(2)8/h3-6H2,1-2H3

InChI key

CATSNJVOTSVZJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Heptanone, a volatile organic compound, belongs to a class of odorous ketones.[1] The impact of 2-heptanone on the behavioral responses of worker honeybees has been reported.[2] The kinetic of reduction of 2-heptanone by samarium iodide in the presence of hexamethylphosphoramide has been analyzed by gas chromatography (GC).[3]

Application

2-Heptanone has been used as a standard odorous compound in a study to understand the odor responses in basiconic, intermediate and trichoid olfactory receptor neurons of Drosophila melanogaster.[4]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ontogeny of the fine structure of the mandibular glands of the honeybee (Apis mellifera L.) workers and the pheromonal activity of 2-heptanone.
Vallet A, et al.
Journal of Insect Physiology, 37(11), 789-804 (1991)
Rate study of reduction of 2-heptanone with samarium iodide by gas chromatography.
Fuh MRS, et al.
Talanta, 46(5), 861-866 (1998)
Oxidation of 2-heptanone in air by a DBD-type plasma generated within a honeycomb monolith supported Pt-based catalyst.
Ayrault C, et al.
Catalysis Today, 89(1), 75-81 (2004)
Alina Silvia Chiper et al.
The journal of physical chemistry. A, 114(1), 397-407 (2009-12-05)
The products of 2-heptanone conversion by dielectric barrier discharge plasma are analyzed under different conditions: alternating current (ac) or pulsed mode of excitation, variable energy, variable composition of the carrier gas. The efficiency of the conversion is higher using a
Ana G Gutiérrez-García et al.
Physiology & behavior, 91(1), 166-172 (2007-04-06)
The present study was aimed to determine whether the urine from donor rats, which were physically stressed (UD-PS) by unavoidable electric footshocks, produces despair in receptor partner rats (RP) in the long-term. For each trial, an RP rat was placed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service