Skip to Content
Merck
All Photos(1)

Key Documents

41640

Sigma-Aldrich

Dimethyl sulfoxide

puriss. p.a., ACS reagent, ≥99.9% (GC)

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
189 °C (lit.)
vapor pressure:
0.42 mmHg ( 20 °C)

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

technique(s)

GC/GC: suitable

impurities

≤0.001% free acid (as CH3COOH)
≤0.1% water

evapn. residue

≤0.005%

refractive index

n20/D 1.479 (lit.)
n20/D 1.479

bp

189 °C (lit.)

mp

16-19 °C (lit.)
18-20 °C

density

1.10 g/mL (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.5 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.5 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfoxide (DMSO) is a polar solvent. DMSO activated by electrophile (oxalyl chloride) has been used to oxidize various alcohols (primary, secondary, allylic, benzylic, hindered and bicyclic). Carbonyl compounds are formed as reaction products. It has been proposed as an alternate solvent for methyl cellosolve for use in ninhydrin reaction, due to its high stability.

Application


  • Modification of gelatin by reaction with 1,6-diisocyanatohexane.: This study focuses on the modification of biopolymers using chemical agents, which could be relevant in contexts where DMSO is used as a reaction solvent ( Bertoldo et al., 2007).

  • Characterization of O-acetyl-(4-O-methylglucurono)xylan isolated from birch and beech.: This research characterizes specific wood-derived xylans, potentially enhancing understanding of such biopolymers when dissolved in DMSO for analytical studies ( Teleman et al., 2002).

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Other Notes

For information on dimethyl sulfoxide miscibility, please visit the following link:
Dimethyl Sulfoxide Miscibility/Immiscibility Table
Use as reagent in synthesis

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.J. Mancuso et al.
Synthesis, 165-165 (1981)
Birgit Dreier et al.
Methods in molecular biology (Clifton, N.J.), 805, 261-286 (2011-11-19)
Ribosome display has proven to be a powerful in vitro selection and evolution method for generating high-affinity binders from libraries of folded proteins. It has been successfully applied to single-chain Fv fragments of antibodies and alternative scaffolds, such as Designed
Oxidation of alcohols by ?activated? dimethyl sulfoxide. A preparative, steric and mechanistic study.
Omura K and Swern D.
Tetrahedron, 34(11), 1651-1660 (1978)
Amino acid analysis: aqueous dimethyl sulfoxide as solvent for the ninhydrin reaction.
S Moore
The Journal of biological chemistry, 243(23), 6281-6283 (1968-12-10)
A RAPID PERMETHYLATION OF GLYCOLIPID, AND POLYSACCHARIDE CATALYZED BY METHYLSULFINYL CARBANION IN DIMETHYL SULFOXIDE.
S HAKOMORI
Journal of biochemistry, 55, 205-208 (1964-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service