Skip to Content
Merck
All Photos(3)

Key Documents

342386

Sigma-Aldrich

Barium hydroxide monohydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(OH)2 · H2O
CAS Number:
Molecular Weight:
189.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

vapor pressure

300 hPa ( 78 °C)

Quality Level

Assay

98%

form

powder, crystals or chunks

density

3.743 g/mL at 25 °C (lit.)

SMILES string

O.O[Ba]O

InChI

1S/Ba.3H2O/h;3*1H2/q+2;;;/p-2

InChI key

GKQTUHKAQKWLIN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M F Byford
The Biochemical journal, 280 ( Pt 1), 261-265 (1991-11-15)
The beta-elimination of phosphoserine residues by dilute alkali is catalysed by the presence of group II metal ions. The use of 0.1 M-Ba (OH)2 catalysed the rate of beta-elimination of phosphoserine by more than two orders of magnitude compared with
K Freeman et al.
Journal of endodontics, 20(6), 272-275 (1994-06-01)
To study the ability of calcium hydroxide to promote hard tissue repair, Alza Alzet Osmotic Pumps, implanted in Sprague-Dawley rats, were used to deliver either calcium hydroxide and glycerol, barium hydroxide and glycerol, tetracycline and glycerol, or glycerol only to
D I Segura et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 69(1), 1-6 (1994-01-01)
Two different histochemical methods for blocking the guanidinium group of arginine in formalin fixed, paraffin embedded material are reported here. One procedure uses diacetyl with short incubation times and high pH, the other uses the trimer of the diacetyl reagent
L Toniolo et al.
Annali di chimica, 91(11-12), 813-821 (2002-02-12)
The Arch of Peace, by Luigi Cagnola, is one of the most famous neoclassical monuments in Milan. It has been subjected to conservative intervention in 1998. In the present paper the efficacy of the consolidation by means of barium hydroxide
V V Onoprienko et al.
Bioorganicheskaia khimiia, 26(6), 403-410 (2000-08-03)
Peptides Boc-Ala-Asn/Gln-OH and Boc-Asn/Gln-Ala-OH were saponified with barium hydroxide to corresponding Asp/Glu-containing peptides. Under the conditions of saponification, Boc-Asn-Ala-OH additionally afforded Boc-Asp-OH, isopeptide Boc-Asp(Ala)-OH, and Boc-NHSuc > Ala-OH, with the third being the key intermediate in these transformations. Boc-Asp(OMe)-Ala-OMe underwent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service