Skip to Content
Merck
All Photos(1)

Key Documents

342270

Sigma-Aldrich

Carbon disulfide

ReagentPlus®, low benzene, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
Beilstein:
1098293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
reagent
bp:
46 °C (lit.)
vapor pressure:
5.83 psi

grade

reagent

Quality Level

vapor density

2.67 (vs air)

vapor pressure

5.83 psi

product line

ReagentPlus®

Assay

≥99.9%

form

liquid

autoignition temp.

212 °F

purified by

glass distillation

expl. lim.

50 %

dilution

(for general lab use)

impurities

<0.05% water
<1 ppm Benzene

evapn. residue

<0.0003%

refractive index

n20/D 1.627 (lit.)

bp

46 °C (lit.)

mp

−112-−111 °C (lit.)

density

1.266 g/mL at 25 °C (lit.)

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Carbon disulfide can be used as a solvent:
  • To synthesize hydroxynaphthyl ketones via Friedel-Crafts acylation and demethylation.
  • In the regioselective bromination of binaphthols.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loric Büchler et al.
Forensic science international, 318, 110590-110590 (2020-12-06)
Arsonists may use ignitable liquids to start, accelerate and amplify fires. The sampling of volatiles present on the hands of suspected arsonists is therefore sometimes carried out in the course of the investigation of (possible) deliberate fires. Several collection protocols
Mark B van Eldijk et al.
Chemical communications (Cambridge, England), 49(71), 7770-7772 (2013-06-19)
CS2 hydrolase, a zinc-dependent enzyme that converts carbon disulfide to carbon dioxide and hydrogen sulfide, exists as a mixture of octameric ring and hexadecameric catenane forms in solution. A combination of size exclusion chromatography, multi-angle laser light scattering, and mass
Continuous direct spinning of fibers of single-walled carbon nanotubes with metallic chirality.
Rajyashree M Sundaram et al.
Advanced materials (Deerfield Beach, Fla.), 23(43), 5064-5068 (2011-10-11)
Huan Liu et al.
Journal of hazardous materials, 235-236, 298-306 (2012-08-21)
Chemical conditioners are often used to enhance sewage sludge dewaterability through altering sludge properties and flocs structure, both affect odorous compounds emissions not only during sludge conditioning but also in subsequent sludge disposal. This study was to investigate emission characteristics
R W Wood
Neurobehavioral toxicology and teratology, 3(4), 397-405 (1981-01-01)
Carbon disulfide is an exceptionally potent neurotoxicant. In industrial settings its effects are pernicious, producing symptoms and behavioral alterations in the absence of frank poisoning; affective disorders are common, and exposure is associated with an elevated incidence of suicide. With

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service