334286
Molecular sieves, 3 Å
pellets, ~1.6 mm
Synonym(s):
Molecular Sieve UOP Type 3Å
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
KnNa12-n[(AlO2)12(SiO2)12] · xH2O
CAS Number:
MDL number:
UNSPSC Code:
23201100
eCl@ss:
32110204
NACRES:
NB.24
Recommended Products
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Molecular sieves, 3 Å is a porous solid that can be used in desiccators and drying tubes to dry gases and absolute solvents. It also changes its color from brown to yellowish at a water uptake of approximately 7-10 g/100 g molecular sieve.
Features and Benefits
- Easy to use
- Chemically inert
- Non-toxic
- Dried liquids can be decanted
- High adsorption capacity
- Selective adsorption
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Joshua A Snyder et al.
The journal of physical chemistry. A, 121(27), 5136-5146 (2017-06-20)
Boron-nitrogen doping of polyaromatic hydrocarbons (PAH), such as borazine-core hexabenzocoronene, presents possibilities for tuning the properties of organic electronics and nanographene materials while preserving structural characteristics of pure hydrocarbons. Previous photochemical studies have demonstrated extension of a borazine-core PAH network
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service