Skip to Content
Merck
All Photos(1)

Key Documents

320196

Sigma-Aldrich

tert-Butyl methyl ether

reagent grade, 98%

Synonym(s):

MTBE, Methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCH3
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730942
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.07
grade:
reagent grade
Assay:
98%
bp:
55-56 °C (lit.)
vapor pressure:
4.05 psi ( 20 °C)

grade

reagent grade

Quality Level

vapor density

3.1 (vs air)

vapor pressure

4.05 psi ( 20 °C)

Assay

98%

form

liquid

autoignition temp.

705 °F

expl. lim.

15.1 %

refractive index

n20/D 1.369 (lit.)

bp

55-56 °C (lit.)

mp

-110 °C

density

0.74 g/mL at 25 °C (lit.)

SMILES string

COC(C)(C)C

InChI

1S/C5H12O/c1-5(2,3)6-4/h1-4H3

InChI key

BZLVMXJERCGZMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of methyl tert-butyl ether (MTBE) with Nafion.
Lien HL and Zhang WX
Journal of Hazardous Materials, 144(1), 194-199 (2007)
J R Hanson et al.
Applied and environmental microbiology, 65(11), 4788-4792 (1999-11-05)
A bacterial strain, PM1, which is able to utilize methyl tert-butyl ether (MTBE) as its sole carbon and energy source, was isolated from a mixed microbial consortium in a compost biofilter capable of degrading MTBE. Initial linear rates of MTBE
Kun-Chang Huang et al.
Chemosphere, 49(4), 413-420 (2002-10-09)
The kinetics of heat-assisted persulfate oxidation of methyl tert-butyl ether (MTBE) in aqueous solutions at various pH, temperature, oxidant concentration and ionic strength levels was studied. The MTBE degradation was found to follow a pseudo-first-order decay model. The pseudo-first-order rate
R J Steffan et al.
Applied and environmental microbiology, 63(11), 4216-4222 (1997-11-15)
Several propane-oxidizing bacteria were tested for their ability to degrade gasoline oxygenates, including methyl tert-butyl ether (MTBE), ethyl tert-butyl ether (ETBE), and tert-amyl methyl ether (TAME). Both a laboratory strain and natural isolates were able to degrade each compound after
Sarah K Abbott et al.
Lipids, 48(3), 307-318 (2013-01-29)
We have developed a protocol suitable for high-throughput lipidomic analysis of human brain samples. The traditional Folch extraction (using chloroform and glass-glass homogenization) was compared to a high-throughput method combining methyl-tert-butyl ether (MTBE) extraction with mechanical homogenization utilizing ceramic beads.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service