Skip to Content
Merck
All Photos(3)

Key Documents

310484

Sigma-Aldrich

Potassium nitrite

ACS reagent, ≥96.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO2
CAS Number:
Molecular Weight:
85.10
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
powder or crystals

grade

ACS reagent

Assay

≥96.0%

form

powder or crystals

impurities

≤0.01% insolubles

pH

7-10 (25 °C, 50 g/L)

mp

350 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.03%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.005%
Fe: ≤0.001%
Mg: ≤0.002%
Na: ≤0.5%
heavy metals (as Pb): ≤0.001%

SMILES string

[K+].[O-]N=O

InChI

1S/K.HNO2/c;2-1-3/h;(H,2,3)/q+1;/p-1

InChI key

BXNHTSHTPBPRFX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium nitrite is an ionic salt that can be used for a variety of chemical reactions, including configuration inversion, substitution of amines via disulfonylamides, nucleophilic aromatic nitration, diazotization, nitrosation, and nucleoside nitration.

Application

Potassium nitrite can be used as:
  • A nucleophile for the nitrosation of diclofenac and diphenylamine into their corresponding N-nitroso derivatives.
  • A nitrous acid source for the diazotization of arylamines.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the crystal structure of the trigonal (and cubic) phase of potassium nitrite.
Solbakk JK and Str?mme KO.
Acta Chemica Scandinavica, 23(1), 20-20 (1969)
Potassium Nitrite
Moriarty R, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis (2001)
Ledgard J.
The Preparatory Manual of Explosives., 167-167 (2014)
Reactions of 4-nitrophthalic anhydride with potassium fluoride and potassium nitrite.
Markezich RL, et al.
The Journal of Organic Chemistry, 42(21), 3435-3436 (1977)
Biotic nitrosation of diclofenac in a soil aquifer system (Katari watershed, Bolivia)
Chiron S and Duwig C
The Science of the Total Environment, 565 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service