Skip to Content
Merck
All Photos(1)

Key Documents

308250

Sigma-Aldrich

tert-Butanol

suitable for HPLC, ≥99.5%

Synonym(s):

2-Methyl-2-propanol, tert-Butyl alcohol, Trimethyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COH
CAS Number:
Molecular Weight:
74.12
Beilstein:
906698
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.5 (vs air)

vapor pressure

31 mmHg ( 20 °C)
44 mmHg ( 26 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

896 °F

expl. lim.

8 %

technique(s)

HPLC: suitable

impurities

<0.050% water

evapn. residue

<0.0005%

refractive index

n20/D 1.387 (lit.)

pH

7 (20 °C)

bp

83 °C (lit.)

mp

23-26 °C (lit.)

density

0.775 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 215 nm Amax: 1.00
λ: 230 nm Amax: 0.50
λ: 250 nm Amax: 0.20
λ: 300-350 nm Amax: 0.01

application(s)

food and beverages

SMILES string

CC(C)(C)O

InChI

1S/C4H10O/c1-4(2,3)5/h5H,1-3H3

InChI key

DKGAVHZHDRPRBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This solvent is glass distilled, submicron filtered and subjected to rigorous specification testing to provide lot-to-lot consistency.

Application

Designed for use with HPLC instrumentation and organic synthesis applications.
Used in the analysis of volatile organic compounds (VOCs) in alcoholic beverages by GC coupled to Flame Ionization Detector (FID).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The photolysis of potassium peroxodisulphate in aqueous solution in the presence of tert-butanol: a simple actinometer for 254nm radiation.
Mark G, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 55(2), 157-160 (1990)
Alkylation of p-cresol with tert-butanol catalyzed by heteropoly acid supported on zirconia catalyst.
Devassy BM, et al.
J. Mol. Catal. A: Chem., 210(1), 125-130 (2004)
Jaime Salazar et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 81(1), 82-90 (2012-01-12)
Nanosizing is a non-specific approach to improve the oral bioavailability of poorly soluble drugs. The decreased particle size of these compounds results in an increase in surface area. The outcome is an increased rate of dissolution, which can lead to
Ee Ling Yong et al.
Water research, 46(6), 1990-1998 (2012-02-14)
Ozonation is widely employed in water treatment to purify water. The R(ct) concept, which is defined as the ratio of OH exposure to ozone exposure, has been commonly used to quantify the OH concentration generating from ozone decomposition and model
Arnau Bassegoda et al.
Applied microbiology and biotechnology, 97(19), 8559-8568 (2013-01-22)
Rhodococci are highly adaptable bacteria, capable to degrade or transform a large number of organic compounds, including recalcitrant or toxic products. However, little information is available on the lipases of the genus Rhodococcus, except for LipR, the first lipase isolated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service