Skip to Content
Merck
All Photos(1)

Key Documents

25044

Sigma-Aldrich

Zinc acetate dihydrate

puriss., E 650, 99-102%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(CH3COO)2 · 2H2O
CAS Number:
Molecular Weight:
219.51
Beilstein:
3732513
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals

grade

puriss.

Assay

99-102%

form

powder or crystals

quality

E 650

reaction suitability

reagent type: catalyst
core: zinc

impurities

organic volatile impurities, complies (GC)
≤0.005% insoluble matter in CH3COOH
≤0.2% alkalis/earth alkalis

pH

6-8 (20 °C, 5%)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤3 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Na: ≤50 mg/kg
Pb: ≤20 mg/kg

SMILES string

O.O.CC(=O)O[Zn]OC(C)=O

InChI

1S/2C2H4O2.2H2O.Zn/c2*1-2(3)4;;;/h2*1H3,(H,3,4);2*1H2;/q;;;;+2/p-2

InChI key

BEAZKUGSCHFXIQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of zinc acetate dihydrate, Zn(CH3COO)2.2H2O.
Van Niekerk JN, et al.
Acta Crystallographica, 6(8), 720-720 (1953)
Synthesis of ZnO nanowires by thermal decomposition of zinc acetate dihydrate.
Lin CC and Li YY.
Materials Chemistry and Physics, 113(1), 102-106 (2009)
Synthesis and optical properties of nanocrystalline ZnO powders by a simple method using zinc acetate dihydrate and poly (vinyl pyrrolidone).
Maensiri S, et al.
Journal of Crystal Growth, 289(1), 102-106 (2006)
Alim Solmaz et al.
Langmuir : the ACS journal of surfaces and colloids, 28(28), 10592-10596 (2012-06-15)
Tapping mode atomic force microscopy (TM-AFM) enables mapping of chemical composition at the nanoscale by taking advantage of the variation in phase angle shift arising from an embedded second phase. We demonstrate that phase contrast can be attributed to the
Tarek Alammar et al.
ChemSusChem, 4(12), 1796-1804 (2011-12-14)
Ultrasound synthesis of zinc oxide from zinc acetate and sodium hydroxide in ionic liquids (ILs) is a fast, facile, and effective, yet highly morphology- and size-selective route to zinc oxide nanostructures of various dimensionalities. No additional organic solvents, water, surfactants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service