243353
Barium sulfate
ReagentPlus®, 99%
Synonym(s):
Baryte
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Linear Formula:
BaSO4
CAS Number:
Molecular Weight:
233.39
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.55
Assay:
99%
form:
powder
Recommended Products
Quality Level
product line
ReagentPlus®
Assay
99%
form
powder
pH
7
SMILES string
[Ba++].[O-]S([O-])(=O)=O
InChI
1S/Ba.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
InChI key
TZCXTZWJZNENPQ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Barium sulphate is widely employed as an inorganic filler. Its crystals belong to the rhombic crystal system. The rhombic crystals transforms to monoclinic form at 1148°C and eventually it decomposes to barium oxide, sulfur dioxide and oxygen at temperatures above 1400°C.
Application
Barium sulfate may be used in the following studies:
- Qualitative and quantitative analysis of Ba2+ and SO4-2 ions.
- Preparation of sulfur dioxide.
- Preparation of hybrid organic-inorganic material.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Improved method for the preparation of sulfur dioxide from barium sulfate for isotope ratio studies.
Bailey SA and Smith JW.
Analytical Chemistry, 44(8), 1542-1543 (1972)
Ionic liquid-functionalized crystals of barium sulfate: A hybrid organic-inorganic material with tuned hydrophilicity and solid-liquid behavior.
Kowacz M, et al.
Materials Chemistry and Physics, 160, 308-314 (2015)
Gregory S Cooper et al.
Cancer, 119(10), 1800-1807 (2013-02-26)
Professional society guidelines recommend follow-up colonoscopy for patients with resected colonic adenomas. However, adherence to guideline recommendations in routine clinical practice has not been well characterized. The authors used a population-based sample of Medicare beneficiaries to identify all patients aged
Steve Halligan et al.
Lancet (London, England), 381(9873), 1185-1193 (2013-02-19)
Barium enema (BE) is widely available for diagnosis of colorectal cancer despite concerns about its accuracy and acceptability. Computed tomographic colonography (CTC) might be a more sensitive and acceptable alternative. We aimed to compare CTC and BE for diagnosis of
Nam P Nguyen et al.
PloS one, 8(3), e56290-e56290 (2013-03-19)
The study aims to assess the feasibility of Tomotherapy-based image-guided radiotherapy (IGRT) to reduce the aspiration risk in patients with non-laryngeal and non-hypopharyngeal cancer. A retrospective review of 48 patients undergoing radiation for non-laryngeal and non-hypopharyngeal head and neck cancers
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service