Skip to Content
Merck
All Photos(5)

Documents

224316

Sigma-Aldrich

Zirconyl chloride octahydrate

greener alternative

reagent grade, 98%

Synonym(s):

Zirconium(IV) oxide chloride, Zirconium(IV) oxychloride octahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrOCl2 · 8H2O
CAS Number:
Molecular Weight:
322.25
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

grade

reagent grade

Quality Level

Assay

98%

form

powder or crystals

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Zr](Cl)=O

InChI

1S/2ClH.8H2O.O.Zr/h2*1H;8*1H2;;/q;;;;;;;;;;;+2/p-2

InChI key

IZYKRMGFEBSOSO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is used as a potential green catalyst. Click here for more information.

Application

Building block for a new series of mixed zirconium phosphonates which may have uses in catalysis, ion exchange or photophysics.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High surface area solids obtained by reaction of montmorillonite with zirconyl chloride.
Yamanaka S and Brindley GW.
Clays and Clay Minerals, 27(2), 119-119 (1979)
Preparation and properties of zirconia coatings from aquo-organic solutions of zirconyl chloride octahydrate.
Atta AP, et al.
Thin Solid Films, 197(1), 187-193 (1997)
The crystal structure of zirconyl chloride octahydrate and zirconyl bromide octahydrate.
Clearfield A and Vaughan PA.
Acta Crystallographica, 9(7), 555-558 (1956)
Refinement of the crystal structure of zirconyl chloride octahydrate.
Mak TCW.
Canadian Journal of Chemistry, 46(22), 3491-3497 (1968)
Anne C Roulin et al.
PloS one, 9(4), e94098-e94098 (2014-04-12)
The European earwig (Forficula auricularia) is an established system for studies of sexual selection, social interactions and the evolution of parental care. Despite its scientific interest, little knowledge exists about the species at the genomic level, limiting the scope of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service