Skip to Content
Merck
All Photos(3)

Key Documents

221325

Sigma-Aldrich

Potassium sulfate

ACS reagent, ≥99.0%, powder or crystals

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

Assay

≥99.0%

form

powder or crystals

impurities

≤0.01% insolubles
≤5 ppm N compounds

pH

5.5-5.8 (25 °C, 5%)

anion traces

chloride (Cl-): ≤0.001%

cation traces

Ca: ≤0.01%
Fe: ≤5 ppm
Mg: ≤0.005%
Na: ≤0.02%
heavy metals: ≤5 ppm (by ICP)

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium sulfate is a water-soluble inorganic salt that is commonly used in fertilizers. It can be prepared by reacting phosphogypsum and potassium chloride. K2SO4 can form needle-like mullite particles on heating with aluminum sulfate and silicon dioxide. Its application to the soil has shown to minimize bronzing of rice plants.

Application

Potassium sulfate may be used as a sulfonating agent in the preparation of 4′-dibenzo-18-crown-6-sulfonic acid via sulfonation of dibenzo-18-crown-6.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfonation of benzocrown ethers by potassium sulfate in polyphosphoric acid.
Grebenyuk AD, et al.
Chemistry of Heterocyclic Compounds, 37(7), 822-826 (2001)
Eagleson M.
Concise Encyclopedia Chemistry, 890-890 (1994)
Conversion of phosphogypsum to potassium sulfate.
Aagli A, et al.
Journal of Thermal Analysis and Calorimetry, 82(2), 395-399 (2005)
Rice bronzing in Nigeria caused by nutrient imbalances and its control by potassium sulfate application.
Yamauchi M.
Plant and Soil, 117(2), 275-286 (1989)
Synthesis of needlelike mullite particles using potassium sulfate flux.
Hashimoto S and Yamaguchi A.
J. Eur. Ceram. Soc., 20(4), 397-402 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service