Skip to Content
Merck
All Photos(1)

Key Documents

221244

Sigma-Aldrich

Ammonium nitrate

ACS reagent, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4NO3
CAS Number:
Molecular Weight:
80.04
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38060309
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

≥98%

form

powder, crystals or granules

impurities

≤0.005% insolubles

ign. residue

≤0.01%

pH

4.5-6.0 (25 °C, 5%)

bp

210 °C (lit.)

mp

169 °C (lit.)

anion traces

chloride (Cl-): ≤5 ppm
nitrite (NO2-): passes test (lim. ~5 ppm)
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.002%

cation traces

Fe: ≤2 ppm
heavy metals (as Pb): ≤5 ppm

SMILES string

N.O[N+]([O-])=O

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3

InChI key

PRORZGWHZXZQMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium nitrate is a major component in nitrogen fertilizers explosives and propellant oxidizer.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Fertilizer Encyclopedia, 46-46 null
Ammonium nitrate: combustion mechanism and the role of additives.
Sinditskii VP, et al.
Propellants, Explosives, Pyrotechnics, 30(4), 269-280 (2005)
Sarah J Benson et al.
Science & justice : journal of the Forensic Science Society, 49(2), 73-80 (2009-07-18)
An evaluation was undertaken to determine if isotope ratio mass spectrometry (IRMS) could assist in the investigation of complex forensic cases by providing a level of discrimination not achievable utilising traditional forensic techniques. The focus of the research was on
Synthesis and characterization of hybrid MCM-41 materials for heavy metal adsorption.
Dimos K, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 126(1), 65-71 (2009)
Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 41(34), 10156-10160 (2012-07-27)
Nanoscale manganese oxides within Faujasite zeolite have been synthesized with a simple method and characterized by scanning electron microscopy, X-ray diffraction spectrometry, N(2) adsorption-desorption isotherms, transmission electron microscopy, and atomic absorption spectroscopy. These oxides showed efficient water oxidizing activity in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service