Skip to Content
Merck
All Photos(3)

Key Documents

216615

Sigma-Aldrich

Potassium chromate

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CrO4
CAS Number:
Molecular Weight:
194.19
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
crystals

grade

ACS reagent

Quality Level

vapor density

6.7 (vs air)

Assay

≥99.0%

form

crystals

impurities

≤0.005% insolubles

pH

8.6-9.8 (25 °C, 5%)

mp

971 °C (lit.)

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.03%

cation traces

Ca: ≤0.005%
Na: ≤0.02%

SMILES string

[K+].[K+].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.2K.4O/q;2*+1;;;2*-1

InChI key

XMXNVYPJWBTAHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium chromate (K2CrO4) is an inorganic solid compound. It is mainly used as an oxidizing agent in the synthesis of organic compound.

Application

Potassium chromate is used as an oxidizing agent:
  • In the conversion of primary and secondary alcohol into aldehydes and ketones using tetrabutylammonium hydrogen sulfate catalyst.

Potassium chromate is also used:
  • To prepare magnetoactive compound by reacting with iron(II) sulfate.
  • As a reagent in the conversion primary allylic and benzylic halides to aldehydes.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zubeyde Bayram-Weston et al.
Journal of neuroscience methods, 265, 81-88 (2015-10-16)
The Golgi-Cox stain is an established method for characterising neuron cell morphology. The method highlights neurite processes of stained cells allowing the complexity of dendritic branching to be measured. Conventional rapid Golgi and Golgi-Cox methods all require fresh impregnation in
Redetermination of the structures of potassium sulphate and potassium chromate: the effect of electrostatic crystal forces upon observed bond lengths.
McGinnety JA.
Acta Crystallographica Section B, Structural Science, 28(9), 2845-2852 (1972)
Comparative Study of Factors Influencing the Action of Corrosion Inhmitors for Mild Steel in Neutral Solution: II. Potassium Chromate.
Mercer AD and Jenkins IR.
British Corrosion Journal, 3(3), 130-135 (1968)
Eagleson M.
Concise Encyclopedia Chemistry, 887-887 (1994)
Kyra T Newmaster et al.
Nature communications, 11(1), 1885-1885 (2020-04-22)
The oxytocin receptor (OTR) plays critical roles in social behavior development. Despite its significance, brain-wide quantitative understanding of OTR expression remains limited in postnatally developing brains. Here, we develop postnatal 3D template brains to register whole brain images with cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service