Skip to Content
Merck
All Photos(3)

Key Documents

216011

Sigma-Aldrich

Ammonium fluoride

ACS reagent, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4F
CAS Number:
Molecular Weight:
37.04
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38050106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

≥98.0%

form

crystals, granules or chunks

impurities

≤0.005% insolubles

ign. residue

≤0.01%

pH

~6 (20 °C)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤5 ppm
heavy metals (as Pb): ≤5 ppm

SMILES string

N.F

InChI

1S/FH.H3N/h1H;1H3

InChI key

LDDQLRUQCUTJBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in a new hydrothermal synthesis of complex flourides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important for solid-state lasers as well as for photoluminescence and magnetic characteristics.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Phase transition in ammonium fluoride.
Stevenson R.
J. Chem. Phys. , 34(1), 346-347 (1961)
Self-organized TiO2 nanotubes prepared in ammonium fluoride containing acetic acid electrolytes.
Tsuchiya H, et al.
Electrochemical Communications, 7(6), 576-580 (2005)
Jae Hoon Bong et al.
Nanoscale, 6(15), 8503-8508 (2014-06-21)
We report a post-synthetic n-doping method for chemical-vapor-deposition (CVD) grown graphene using wet chemical processing. An ammonium fluoride solution was found effective in converting pristine hole doping into electron doping in addition to the mobility improvement of charge carriers. We
Xun, X. et al.
Chemistry of Materials, 9, 2966-2966 (1997)
On the Etching of Silicon by Oxidants in Ammonium Fluoride Solutions A Mechanistic Study.
Gerischer H and Lubke M.
Journal of the Electrochemical Society, 135(11), 2782-2786 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service