Skip to Content
Merck
All Photos(4)

Key Documents

215929

Sigma-Aldrich

Potassium iodate

ACS reagent, 99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KIO3
CAS Number:
Molecular Weight:
214.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder

grade

ACS reagent

Assay

99.4-100.4% (ACS specification)
99.5%

form

powder

impurities

≤0.005% N compounds
≤0.005% insolubles

pH

5.0-8.0 (25 °C, 5%)

mp

560 °C (lit.)

density

3.93 g/mL at 25 °C (lit.)

anion traces

chloride, bromide (as Cl)-: ≤0.01%
iodide (I-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
Na: ≤0.005%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[K+].[O-]I(=O)=O

InChI

1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

JLKDVMWYMMLWTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

KIO3 can be used as a substitute of KI in radiation protection. A kinetic study of thermal degradation of KIO3 by γ-rays suggests that rate of decomposition increases while activation energy decreases upon irradiation.

Application

Potassium iodate (KIO3) may be used as an oxidizing agent in the estimation of hydrolyzable tannins (gallotannins and ellagitannins) in plant samples. It may also be used in the preparation of polyaniline, poly(o-anisidine), poly[aniline-co-(o-anisidine) and TiO2 nanoparticles.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal decomposition kinetics of potassium iodate.
Muraleedharan K.
Journal of Thermal Analysis and Calorimetry, 114(2), 491-496 (2013)
Mechanistic study of osmium (VIII) promoted oxidation of crotonic acid by aqueous alkaline solution of potassium iodate
Singh B, et al.
Indian Journal of Chemistry, 54, 1387-1393 (2015)
Synthesis and properties of polyaniline, poly (o-anisidine), and poly [aniline-co-(o-anisidine)] using potassium iodate oxidizing agent
Neetika M, et al.
High Performance Polymers (2016)
Yawen Wang et al.
Journal of colloid and interface science, 430, 31-39 (2014-07-07)
In this paper, we report a novel polyol process to synthesize highly water-dispersible anatase titanium dioxide (TiO2) nanoparticles (∼5 nm) by the introduction of inorganic oxidizing agent--KIO3. The obtained TiO2 nanoparticles are well dispersible in water at pH≥5.0 and the
Paul W Hartzfeld et al.
Journal of agricultural and food chemistry, 50(7), 1785-1790 (2002-03-21)
A widely used method for analyzing hydrolyzable tannins afer reaction with KIO(3) has been modified to include a methanolysis step followed by oxidation with KIO(3). In the new method, hydrolyzable tannins (gallotannins and ellagitannins) are reacted at 85 degrees C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service