Skip to Content
Merck
All Photos(2)

Key Documents

215783

Sigma-Aldrich

Antimony(III) chloride

ReagentPlus®, 99%

Synonym(s):

Antimony trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbCl3
CAS Number:
Molecular Weight:
228.12
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent
form:
crystals or chunks
Pricing and availability is not currently available.

grade

reagent

Quality Level

vapor density

7.9 (vs air)

vapor pressure

1 mmHg ( 49 °C)

product line

ReagentPlus®

Assay

99%

form

crystals or chunks

reaction suitability

reagent type: catalyst
core: antimony

mp

73.4 °C (lit.)

SMILES string

Cl[Sb](Cl)Cl

InChI

1S/3ClH.Sb/h3*1H;/q;;;+3/p-3

InChI key

FAPDDOBMIUGHIN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application


  • Syntheses and characterization of novel antimony (III) and bismuth (III) derivatives containing β-enamino ester along with antimicrobial evaluation, DFT calculation, and cytotoxic study.: This research focuses on the synthesis and characterization of novel antimony(III) chloride derivatives. The study evaluates their antimicrobial properties, conducts Density Functional Theory (DFT) calculations, and assesses cytotoxicity. The findings suggest significant potential for antimony(III) chloride in developing new antimicrobial agents and understanding their biological interactions (Sangwan et al., 2024).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kuan-Ting Chen et al.
ACS nano, 14(9), 11648-11661 (2020-09-05)
The development of high-performance potassium ion battery (KIB) electrodes requires a nanoengineering design aimed at optimizing the construction of active material/buffer material nanocomposites. These nanocomposites will alleviate the stress resulting from large volume changes induced by K+ ion insertion/extraction and
Synthesis, structural characterization and cytotoxicity of the antimony (III) chloride complex with N, N-dicyclohexyldithiooxamide.
Ozturk II, et al.
Polyhedron, 52, 1403-1410 (2013)
Nucleic acid related compounds. 79. Efficient conversions of thioethers to. alpha.-fluoro thioethers with DAST or DAST/antimony (III) chloride.
Robins MJ and Wnuk SF.
The Journal of Organic Chemistry, 58(15), 3800-3800 (1993)
Antimony (III) chloride-catalyzed ring opening of epoxides with anilines.
Singh MC and Peddinti RK.
Tetrahedron Letters, 48(41), 7354-7357 (2007)
Antimony (III) chloride-catalysed Biginelli reaction: a versatile method for the synthesis of dihydropyrimidinones through a different reaction mechanism
Cepanec I, et al.
Tetrahedron, 63(48), 11822-11827 (2007)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service