Skip to Content
Merck
All Photos(3)

Documents

16201

Sigma-Aldrich

Tannic acid

puriss., powder

Synonym(s):

Gallotannin, Tannin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C76H52O46
CAS Number:
Molecular Weight:
1701.20
Beilstein:
8186396
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

puriss.

Quality Level

form

powder

autoignition temp.

980 °F

quality

meets analytical specification of USP

impurities

dextrines, gum matters, in accordance
resinous matters, in accordance

ign. residue

≤0.1% (as SO4)

loss

≤9% loss on drying, 105 °C, 2 h

mp

218 °C (lit.)

solubility

H2O: soluble 1 gm in 0.35ml
glycerol: soluble 1gm in 1 ml (warm)
acetone: very soluble
alcohol: freely soluble (Diluted)
alcohol: slightly soluble (Dehydrated)
alcohol: very soluble
benzene: insoluble
carbon disulfide: insoluble
carbon tetrachloride: insoluble
chloroform: insoluble
diethyl ether: insoluble
hexane: insoluble
petroleum ether: insoluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

Oc1cc(cc(O)c1O)C(=O)Oc2cc(cc(O)c2O)C(=O)OC[C@H]3O[C@@H](OC(=O)c4cc(O)c(O)c(OC(=O)c5cc(O)c(O)c(O)c5)c4)[C@H](OC(=O)c6cc(O)c(O)c(OC(=O)c7cc(O)c(O)c(O)c7)c6)[C@@H](OC(=O)c8cc(O)c(O)c(OC(=O)c9cc(O)c(O)c(O)c9)c8)[C@@H]3OC(=O)c%10cc(O)c(O)c(OC(=O)c%11cc(O)c(O)c(O)c%11)c%10

InChI

1S/C76H52O46/c77-32-1-22(2-33(78)53(32)92)67(103)113-47-16-27(11-42(87)58(47)97)66(102)112-21-52-63(119-72(108)28-12-43(88)59(98)48(17-28)114-68(104)23-3-34(79)54(93)35(80)4-23)64(120-73(109)29-13-44(89)60(99)49(18-29)115-69(105)24-5-36(81)55(94)37(82)6-24)65(121-74(110)30-14-45(90)61(100)50(19-30)116-70(106)25-7-38(83)56(95)39(84)8-25)76(118-52)122-75(111)31-15-46(91)62(101)51(20-31)117-71(107)26-9-40(85)57(96)41(86)10-26/h1-20,52,63-65,76-101H,21H2/t52-,63-,64+,65-,76+/m1/s1

InChI key

LRBQNJMCXXYXIU-PPKXGCFTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tannic acid is the simplest hydrolyzable tannin. The chemical structure of tannic acid has a glucose molecule in its core connected to 10 galloyls groups via aliphatic ester bonds. TA is one of the most abundant natural polyphenols, which can be colorless to yellow in color, and of astringent taste. It can be sourced from the bark of oak, chestnut, hemlock, mangrove, etc.

Application

Tannic acid has been used:
  • as a reference standard to measure the total hydrolyzable tannins in Oak root bark tannin-methanol extract using a UV/Vis spectrophotometer
  • as a blood-brain barrier (BBB) impermeable, transcytosis-inhibiting drug in spheroid culture medium
  • as a flushing agent to test its effects on metal ions, total organic carbon, and organic matter of soil
  • as an inhibitor of free ovalbumin (OVA) uptake by the endocytic mechanism on dendritic cells

Biochem/physiol Actions

Tannic acid has many pharmacological properties such as antioxidant, anti-inflammatory, antimicrobial, and anti-tumor effects. It also exerts anti-aging, wound healing, and neuroprotective activities. TA is traditionally used as a tanning agent in the food, leather, adhesive, and pharmaceutical industries. It is a natural crosslinker that is useful in building polymeric networks and can be used to design biomedical hydrogels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

390.2 °F

Flash Point(C)

199 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tannic acid: a crosslinker leading to versatile functional polymeric networks: a review.
Chen, et al.
Royal Society of Chemistry Advances, 12, 7689-7711 (2022)
Wang Jing et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 154, 113561-113561 (2022-08-28)
In recent years, increasing attention has been paid to the pharmacological efficacy of tannins. Tannic acid (TA), the simplest hydrolysable tannin that has been approved by the FDA as a safe food additive, is one of the most important components
Yanyan Sun et al.
Anti-cancer drugs, 23(9), 979-990 (2012-07-13)
Tannic acid (TA) has been associated with anticancer functions in multiple tumor types both in vitro and in vivo. However, its effect on ovarian carcinoma cells has not been investigated, and its underlying anticancer mechanism(s) remain unclear. In this study
W A Anderson et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 23(4), 295-302 (1975-04-01)
Endogenous mannary gland peroxidase in acinar cells of prelactating and lactating rats is revealed in tannic acid-formaldehyde-glutaraldehyde-fixed tissue by means of the standard diaminobenzidine procedure. Diaminobenzidine cytochemical reaction product is present in perinuclear cisternae, in the granular endoplasmic reticulum and
R C Srivastava et al.
Cancer letters, 153(1-2), 1-5 (2000-04-26)
The deleterious effects of excessive release of nitric oxide (NO) have been implicated in the tissue damage and inflammation. In this study, the effect of various flavonoids and other oxidant scavenging chemical agents have been studied for their ability to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service