Skip to Content
Merck
All Photos(1)

Key Documents

13568

Sigma-Aldrich

Sodium carbonate monohydrate

puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, 99.5-100.5% (ex dried subst.)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3 · H2O
CAS Number:
Molecular Weight:
124.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.5-100.5% (ex dried subst.)
form:
powder or crystals

grade

puriss.

Quality Level

Assay

99.5-100.5% (ex dried subst.)

form

powder or crystals

quality

meets analytical specification of Ph. Eur., BP, NF, FCC, E500

impurities

alkali hydroxides and-bicarbonates, complies
organic volatile impurities, complies (GC)
residual solvents, complies

loss

12-15% loss on drying, 300 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Hg: ≤1 mg/kg
K: ≤300 mg/kg
Pb: ≤4 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution

SMILES string

O.[Na+].[Na+].[O-]C([O-])=O

InChI

1S/CH2O3.2Na.H2O/c2-1(3)4;;;/h(H2,2,3,4);;;1H2/q;2*+1;/p-2

InChI key

MQRJBSHKWOFOGF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Buffer component, may be used for the removal of peripheral membrane proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The quantitative monitoring of mechanochemical reaction between solid L-tartaric acid and sodium carbonate monohydrate by terahertz spectroscopy.
Liu X, et al.
Journal of Physics and Chemistry of Solids, 72(11), 1245-1250 (2011)
Dehydration of sodium carbonate monohydrate with indirect microwave heating.
Seyrankaya A and Ozalp B.
Thermochimica Acta, 448(1), 31-36 (2006)
Feng Gu et al.
Bioresource technology, 124, 299-305 (2012-09-20)
Green liquor consists of sodium carbonate and sodium sulfide and is readily available in any kraft mills. The green liquor pretreatment process for bioethanol production was developed for wood chips. This process uses only proven technology and equipment currently used
Ping Guo et al.
Journal of virological methods, 183(2), 139-146 (2012-05-09)
Preclinical gene therapy studies both in vitro and in vivo require high purity preparations of adeno-associated virus (AAV). Current methods for purification of AAV entail the use of centrifugation over either a CsCl or iodixanol gradient, or the use of
Linfeng Yang et al.
Bioresource technology, 124, 283-291 (2012-09-20)
The effects of sodium carbonate (Na(2)CO(3)) pretreatment on the chemical compositions and enzymatic saccharification of rice straw were investigated. The enzymatic digestibility of rice straw is enhanced after pretreatment since pretreated solids show significant delignification with high sugar availability. During

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service