Skip to Content
Merck
All Photos(1)

Key Documents

13160

Sigma-Aldrich

Benzyl alcohol

puriss. p.a., ACS reagent, ≥99.0% (GC)

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
EC Number:
MDL number:
UNSPSC Code:
12352104
eCl@ss:
39023110
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (GC)
bp:
203-205 °C (lit.)
vapor pressure:
13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

grade

ACS reagent
puriss. p.a.

Quality Level

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

817 °F

expl. lim.

0.34-6.3 %

impurities

≤0.01% benzaldehyde (C6H5CHO)
≤0.02% acetophenone
≤0.02% halogenated compounds (as Cl)
≤0.03% free acid (as benzoic acid)

evapn. residue

≤0.005%

ign. residue

≤0.005% (as SO4)

refractive index

n20/D 1.539 (lit.)
n20/D 1.541

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

solubility

H2O: 33 mg/mL at 20 °C, clear, colorless

density

1.045 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.5 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤2 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl alcohol is an aromatic hydrocarbon commonly employed as solvents and intermediates in cosmetics, pharmaceuticals, and flavor and fragrance products. Due to its moderate toxicity, high polarity, solubility in water and other solvents, low vapor pressure, and pleasantly aromatic scent, benzyl alcohol has gained wide application as a solvent.

Application

Benzyl alcohol is used as a solvent:
  • In the synthesis of ZnO nanoparticles from zinc acetate.
  • Etherification of glycerol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Process intensification: oxidation of benzyl alcohol using a continuous isothermal reactor under microwave irradiation.
Jachuck RJJ, et al.
Green Chemistry, 8(1), 29-33 (2006)
Orazio Fortunato et al.
Frontiers in immunology, 11, 02168-02168 (2020-10-31)
Cancer stem cells (CSCs) are functionally defined as the cell subset with greater potential to initiate and propagate tumors. Within the heterogeneous population of lung CSCs, we previously identified highly disseminating CD133+CXCR4+ cells able to initiate distant metastasis (metastasis initiating
Tongfei A Wang et al.
Neuron, 103(2), 309-322 (2019-06-04)
Body temperature control is essential for survival. In mammals, thermoregulation is mediated by the preoptic area of anterior hypothalamus (POA), with ∼30% of its neurons sensitive to brain temperature change. It is still unknown whether and how these temperature-sensitive neurons
Matteo Caravati et al.
Physical chemistry chemical physics : PCCP, 7(2), 278-285 (2005-01-21)
Selective oxidation of benzyl alcohol to benzaldehyde with molecular oxygen over an alumina-supported palladium catalyst was performed with high rate at about 95% selectivity in supercritical carbon dioxide. The experiments in a continuous flow fixed-bed reactor showed that the pressure
Béatrice Hechler et al.
The Journal of pharmacology and experimental therapeutics, 314(1), 232-243 (2005-03-29)
Our aim was to determine whether the newly described P2X1 antagonist NF449 [4,4',4'',4'''-(carbonylbis(imino-5,1,3-benzenetriylbis(carbonylimino)))tetrakis-benzene-1,3-disulfonic acid octasodium salt] could selectively antagonize the platelet P2X1 receptor and how it affected platelet function. NF449 inhibited alpha,beta-methyleneadenosine 5'-triphosphate-induced shape change (IC50 = 83 +/- 13

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service