Skip to Content
Merck
All Photos(2)

Key Documents

128082

Sigma-Aldrich

2-Ethoxyethanol

ReagentPlus®, 99%

Synonym(s):

Ethyl glycol, Ethylene glycol monoethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1098271
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.1 (vs air)

Quality Level

vapor pressure

3.8 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

460 °F

expl. lim.

14 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.10% water

refractive index

n20/D 1.407 (lit.)

bp

135 °C (lit.)

mp

−90 °C (lit.)

solubility

water: miscible

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CCOCCO

InChI

1S/C4H10O2/c1-2-6-4-3-5/h5H,2-4H2,1H3

InChI key

ZNQVEEAIQZEUHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethoxyethanol is a glycol ether. It has been reported as a precursor for ethoxyacetic acid (EAA). Various physical properties (density, refractive index, viscosity and speed of sound) of its binary mixtures with n-alkanes, 2,2,4-trimethylpentane and cyclohexane have been evaluated. High-pressure vapor-liquid equilibrium (VLE) data of its binary mixtures with CO2 have been reported. It can be used as a solvent in the spectrophotometric determination of iron(III) by using the ligand, N,N-bis(2-hydroxy-5-bromo-benzyl)1,2 diaminopropane (HBDAP).

Application

2-Ethoxyethanol (Ethylene glycol monoethyl ether) has been used in zebrafish embryotoxicity test (ZET). It can act like a co-surfactant in the separation of highly hydrophobic analytes via microemulsion electrokinetic chromatography and solvent-modified micellar electrokinetic chromatography.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Comparison of microemulsion electrokinetic chromatography and solvent-modified micellar electrokinetic chromatography"
Hansen HS, et al.
Journal of Separation Science, 24(8), 643-650 (2001)
"Selective preconcentration, separation and speciation of ferric iron in different samples using N, N'-bis (2-hydroxy-5-bromo-benzyl) 1, 2 diaminopropane"
Kara D and Alkan
Talanta, 55(2), 415-423 (2001)
Density, Viscosity, Refractive Index, and Speed of Sound in Binary Mixtures of 2-Ethoxyethanol with n-Alkanes (C6 to C12), 2, 2, 4-Trimethylpentane, and Cyclohexane in the Temperature Interval 298.15-313.15 K.
Aminabhavi TM and Gopalakrishna B.
Journal of Chemical and Engineering Data, 40(3), 632-641 (1995)
Measurements and correlation of high-pressure VLE of binary CO2-alcohol systems (methanol, ethanol, 2-methoxyethanol and 2-ethoxyethanol).
Joung SN, et al.
Fluid Phase Equilibria, 185(1), 219-230 (2001)
Sanne A B Hermsen et al.
Toxicology in vitro : an international journal published in association with BIBRA, 25(3), 745-753 (2011-01-18)
The zebrafish embryotoxicity test (ZET) is a fast and simple method to study chemical toxicity after exposure of the complete vertebrate embryo during embryogenesis in ovo. We developed a novel quantitative evaluation method to assess the development of the zebrafish

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service