Skip to Content
Merck
All Photos(1)

Key Documents

Y0001576

Oxcarbazepine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

10,11-Dihydro-10-oxo-5h-dibenz[b,f]azepine-5-carboxamide, Oxacarbazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxcarbazepine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1CC2=C(C=CC=C2)N(C(N)=O)C3=CC=CC=C31

InChI

1S/C15H12N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8H,9H2,(H2,16,19)

InChI key

CTRLABGOLIVAIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxcarbazepine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Anticonvulsant, antineuralgic. Inhibits veratrine-induced transmitter release.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Vasudev et al.
The Cochrane database of systematic reviews, (1)(1), CD005171-CD005171 (2008-02-07)
Some studies have suggested that oxcarbazepine has a role in preventing episode recurrence in bipolar affective disorder. This review attempted to investigate the existing evidence from randomised controlled trials for its use in the maintenance treatment of this illness. To
Barbara R Sommer et al.
Expert opinion on drug safety, 6(2), 133-145 (2007-03-21)
Few controlled studies are available to guide the clinician in treating potentially assaultive elderly individuals with psychiatric disorders. Safety concerns limit the use of benzodiazepines and antipsychotic medications in the elderly individual, making anticonvulsants an attractive alternative. This paper reviews
Gaetano Zaccara et al.
Seizure, 22(7), 528-536 (2013-04-30)
Analysis of overall tolerability and neurological adverse effects (AEs) of eslicarbazepine acetate (ESL), lacosamide (LCM) and oxcarbazepine (OXC) from double-blind, placebo-controlled trials. Indirect comparisons of patients withdrawing because of AEs, and the incidence of some vestibulocerebellar AEs between these three
James F Knudsen et al.
Pediatric neurology, 37(2), 134-137 (2007-08-07)
Oxcarbazepine, a carbamazepine analog, was approved for use as an antiepileptic agent in the United States in 2000. A search of the United States Food and Drug Administration's Adverse Event Reporting System identified nine cases of oxcarbazepine-associated angioedema in pediatric
Humberto Saconato et al.
Sao Paulo medical journal = Revista paulista de medicina, 127(3), 150-159 (2009-10-13)
It has been estimated that 50 million people worldwide suffer from epilepsy and around 30% will not achieve adequate control over the disease. The aim was to evaluate the effectiveness of oxcarbazepine for refractory partial or generalized epilepsy. Systematic review.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service