Skip to Content
Merck
All Photos(1)

Key Documents

Y0001426

Carvedilol for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Carvedilol, 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol, BM-14190

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N2O4
CAS Number:
Molecular Weight:
406.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

carvedilol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(COC1=CC=CC2=C1C3=C(C=CC=C3)N2)CNCCOC4=CC=CC=C4OC

InChI

1S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27H,13-16H2,1H3

InChI key

OGHNVEJMJSYVRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carvedilol for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cavedilol is a non-selective β-adrenergic blocker with α1 blocking activity.
Cavedilol is a non-selective β-adrenergic blocker with α1 blocking activity. Carvedilol is used specifically for the treatment of heart failure and high blood pressure. It has been shown to improve left ventricular ejection fraction and may reduce mortality.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2

Target Organs

Liver,spleen,Uterus (including cervix),Adrenal gland

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Randall P Sharp et al.
The Annals of pharmacotherapy, 42(4), 564-571 (2008-03-28)
To examine the evidence regarding the impact of carvedilol on the serum lipid profile. Searches in MEDLINE and International Pharmaceutical Abstracts (1966-December 2007) were conducted. Search terms included carvedilol, cholesterol, lipids, hyperlipidemia, and beta-blockers. Published studies and case reports that
Adrian J Stanley et al.
Journal of hepatology, 61(5), 1014-1019 (2014-06-24)
Rebleeding after an initial oesophageal variceal haemorrhage remains a significant problem despite therapy with band ligation, non-selective β-blockers or a combination of these. Carvedilol is a vasodilating non-selective β-blocker with alpha-1 receptor and calcium channel antagonism. A recent study has
Dhiraj Tripathi et al.
European journal of gastroenterology & hepatology, 22(8), 905-911 (2010-01-23)
Carvedilol is a potent noncardioselective beta-blocker, with weak vasodilating properties because of alpha 1 blockade. A reduction in both intrahepatic and portocollateral resistance contribute to enhanced effects on portal pressure. There are 10 published hemodynamic studies involving 168 patients investigating
Carol Chen-Scarabelli et al.
American journal of cardiovascular drugs : drugs, devices, and other interventions, 12(6), 391-401 (2012-10-16)
β-Adrenergic receptor antagonists (β-blockers) have been recognized for their cardioprotective properties, prompting use of these pharmacologic agents to become more mainstream in acute myocardial infarction (AMI) and congestive heart failure (CHF). Despite their popularity as a class, the ability to
James J DiNicolantonio et al.
The American journal of cardiology, 113(3), 565-569 (2013-12-18)
A systematic review and meta-analysis was performed to evaluate the effects of carvedilol versus metoprolol on the incidence of postoperative atrial fibrillation in patients undergoing coronary artery bypass grafting in randomized controlled trials. Ovid MEDLINE, PubMed, CENTRAL, and Excepta Medica

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service