Skip to Content
Merck
All Photos(1)

Key Documents

Y0001243

Folic acid impurity D

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Pteroic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N6O3
CAS Number:
Molecular Weight:
312.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

folic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC1=NC(=O)C2=NC(CNc3ccc(cc3)C(O)=O)=CNC2=N1

InChI

1S/C14H12N6O3/c15-14-19-11-10(12(21)20-14)18-9(6-17-11)5-16-8-3-1-7(2-4-8)13(22)23/h1-4,6,16H,5H2,(H,22,23)(H3,15,17,19,20,21)

InChI key

JOAQINSXLLMRCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Folic acid impurity D EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ratnesh Jain et al.
International journal of pharmaceutics, 420(1), 147-155 (2011-09-03)
Enhanced intracellular internalization of the anti-cancer active idarubicin (IDA) was achieved through appropriate surface modification of IDA loaded propyl starch nanoparticles. This was conducted by synthesizing pteroic acid modified polyvinyl alcohol (ptPVA) and employing this stabilizer for formulating the said
Michael S Lee et al.
The journal of physical chemistry. B, 112(42), 13411-13417 (2008-09-30)
A comparative analysis is provided of the effect of different solvent models on the calculation of a potential of mean force (PMF) for determining the absolute binding affinity of the small molecule inhibitor pteroic acid bound to ricin toxin A-chain
C E Butterworth et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 181(1), 49-58 (1986-01-01)
"No-blood disease" is a severe, feed-related anemia of channel catfish. Pathological features in advanced cases include almost total absence of circulating red blood cells, hepatic fatty change, megaloblastic arrest of hematopoiesis, and intussusceptions of the small intestine. In view of
Cristina Müller et al.
European journal of nuclear medicine and molecular imaging, 33(9), 1007-1016 (2006-06-10)
The folate receptor (FR) is a valuable tumour marker, since it is frequently overexpressed on various cancer types. The purpose of the present study was to pre-clinically evaluate novel site-specifically modified (99m)Tc(CO)(3) folate (gamma-derivative 4, alpha-derivative 5) and pteroate (6)
R L Stephens et al.
The Journal of pharmacology and experimental therapeutics, 239(3), 627-633 (1986-12-01)
Folic acid (FA) and 5-formyltetrahydrofolic acid (FTHF) have been shown previously to produce a marked stimulation of locomotor activity after bilateral injection into the rat nucleus accumbens. This study was designed to determine whether the hypermotility response produced by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service