Skip to Content
Merck
All Photos(1)

Documents

Y0001094

Aceclofenac for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Aceclofenac, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid carboxymethyl ester;

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13Cl2NO4
CAS Number:
Molecular Weight:
354.18
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

aceclofenac

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl

InChI

1S/C16H13Cl2NO4/c17-11-5-3-6-12(18)16(11)19-13-7-2-1-4-10(13)8-15(22)23-9-14(20)21/h1-7,19H,8-9H2,(H,20,21)

InChI key

MNIPYSSQXLZQLJ-UHFFFAOYSA-N

Gene Information

human ... PTGS2(5743)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Aceclofenac for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Non-steroidal, anti-inflammatory drug (NSAID), with selectivity for COX-2 over COX-1.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Kulthe et al.
Colloids and surfaces. B, Biointerfaces, 88(2), 691-696 (2011-08-25)
Pluronic block copolymers offer affluent phase behavioral characteristics and are extensively investigated for drug delivery applications. Hydrophobic Pluronics produce larger aggregates whereas hydrophilic Pluronics often generate small-sized micelles in aqueous milieu. To overcome the limitations and combine the advantages of
Jhi-Kai Chang et al.
Clinical rheumatology, 32(3), 341-347 (2012-11-30)
This study aimed to determine the safety and efficacy of tramadol 37.5 mg/acetaminophen 325 mg combination tablets (Ultracet®) in patients with ankylosing spondylitis (AS). This was a 12-week, randomized, double-blind, placebo-controlled study. Sixty patients with active AS according to the
Madhusmruti Khandai et al.
Current drug delivery, 9(5), 495-505 (2012-04-24)
The aim of this present research work was to prepare and evaluate alginate microspheres of aceclofenac by ionic gelation method for targeting the drug release in intestinal region and decrease distinct tissue protection in the stomach. This method offers to
R Andrew Moore et al.
The Cochrane database of systematic reviews, (3)(3), CD007588-CD007588 (2009-07-10)
Aceclofenac is the prodrug of the non-steroidal anti-inflammatory drug (NSAID) diclofenac, widely used to treat acute and chronic pain. There are no known systematic reviews of its analgesic efficacy in acute postoperative pain. This review sought to evaluate the efficacy
Kyung Rok Chu et al.
Archives of pharmacal research, 35(7), 1187-1195 (2012-08-07)
This study examined the effects of the particle size of various poorly water-soluble drugs on their dissolution behavior through physicochemical and mathematical analysis. As model drugs, hydrochlorothiazide, aceclofenac, ibuprofen and a discovery candidate were selected. The materials were crystallized using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service