Skip to Content
Merck
All Photos(1)

Key Documents

Y0000773

Lansoprazole

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F3N3O2S
CAS Number:
Molecular Weight:
369.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lansoprazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cc1c(CS(=O)c2nc3ccccc3[nH]2)nccc1OCC(F)(F)F

InChI

1S/C16H14F3N3O2S/c1-10-13(20-7-6-14(10)24-9-16(17,18)19)8-25(23)15-21-11-4-2-3-5-12(11)22-15/h2-7H,8-9H2,1H3,(H,21,22)

InChI key

MJIHNNLFOKEZEW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lansoprazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grace Chai et al.
Pediatrics, 130(1), 23-31 (2012-06-20)
To describe trends in outpatient prescription drug utilization in US children and the changes in major areas of pediatric therapeutic use for the years 2002 through 2010. Large prescription databases (the IMS Vector One: National and Total Patient Tracker) were
D A Peura et al.
Alimentary pharmacology & therapeutics, 37(8), 810-818 (2013-03-05)
Higher body mass index (BMI) is a recognised risk factor for gastro-oesophageal reflux disease (GERD). Data regarding the impact of BMI on proton pump inhibitor (PPI) therapy are conflicting. To assess the impact of BMI on baseline heartburn symptom severity
Nahuai Badiola et al.
PloS one, 8(3), e58837-e58837 (2013-03-23)
A key event in the pathogenesis of Alzheimer's disease (AD) is the accumulation of amyloid-β (Aβ) species in the brain, derived from the sequential cleavage of the amyloid precursor protein (APP) by β- and γ-secretases. Based on a systems biology
Hala H Mosli et al.
The Journal of clinical endocrinology and metabolism, 97(9), E1731-E1735 (2012-06-23)
Chromogranin A (CgA) is used as a generic tumor marker for neuroendocrine tumors. Proton pump inhibitors (PPI) are known to increase CgA, but it is not clear to what extent, and there is little information on how long PPI need
Yun Jeong Lim et al.
Digestion, 86(2), 171-177 (2012-08-22)
Proton pump inhibitors (PPIs) are widely used to prevent nonsteroidal anti-inflammatory drug (NSAID)-induced peptic ulcers. NSAIDs produce small intestinal injury and some PPIs have been reported to protect against NSAID-induced small bowel injury in rats. The aim of this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service