Skip to Content
Merck
All Photos(1)

Key Documents

Y0000723

Thebaine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Paramorphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO3
CAS Number:
Molecular Weight:
311.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

thebaine, oxycodone, codeine

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12Oc3c(OC)ccc4C[C@H]5N(C)CC[C@]1(C5=CC=C2OC)c34

InChI

1S/C19H21NO3/c1-20-9-8-19-12-5-7-15(22-3)18(19)23-17-14(21-2)6-4-11(16(17)19)10-13(12)20/h4-7,13,18H,8-10H2,1-3H3/t13-,18+,19+/m1/s1

InChI key

FQXXSQDCDRQNQE-VMDGZTHMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thebaine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing He et al.
Nature communications, 6, 5943-5943 (2015-01-13)
Alkaloids constitute a large family of natural products possessing diverse biological properties. Their unique and complex structures have inspired numerous innovations in synthetic chemistry. In the realm of late-stage C-H functionalization, alkaloids remain a significant challenge due to the presence
Sándor Berényi et al.
Current medicinal chemistry, 16(25), 3215-3242 (2009-06-25)
The most practical synthetic routes to the preparation of as important pharmaceuticals as oxycodone, naloxone, naltrexone, nalbuphine and buprenorphine have utilized the alkaloid, thebaine, as a starting material. This review intends to focus on chemical transformations of morphinans which resulted
Masako Aragane et al.
Journal of natural medicines, 68(4), 677-685 (2014-06-24)
In May 2011, numerous poppy plants closely resembling Papaver bracteatum Lindl., a type of narcotic plant that is illegal in Japan, were distributed directly from several large flower shops or through online shopping throughout Japan, including the Tokyo Metropolitan area.
Zoe M Smith et al.
Talanta, 130, 221-225 (2014-08-28)
Previous applications of manganese(IV) as a chemiluminescence reagent have required the use of formaldehyde to enhance the emission intensity to analytically useful levels. However, this known human carcinogen (by inhalation) is not ideal for routine application. A wide range of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service