Skip to Content
Merck
All Photos(1)

Key Documents

Y0000593

Chondroitin sulfate sodium (marine)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Chondroitin sulfate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

chondroitin sulfate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O[C@H]1O[C@H]([C@@H]([C@H]([C@H]1O)O[C@@H]2O[C@@H]([C@H]([C@@H]([C@H]2O)O)O)C(=O)O)NC(=O)C)O)O

InChI

1S/C13H21NO15S/c1-2(15)14-3-8(7(19)13(28-11(3)22)29-30(23,24)25)26-12-6(18)4(16)5(17)9(27-12)10(20)21/h3-9,11-13,16-19,22H,1H3,(H,14,15)(H,20,21)(H,23,24,25)/t3-,4+,5+,6-,7-,8-,9+,11-,12-,13-/m1/s1

InChI key

KXKPYJOVDUMHGS-OSRGNVMNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chondroitin sulfate sodium (marine) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trine M Reine et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 61(8), 606-616 (2013-06-13)
Heparan sulfate proteoglycans are hypothesized to contribute to the filtration barrier in kidney glomeruli and the glycocalyx of endothelial cells. To investigate potential changes in proteoglycans in diabetic kidney, we isolated glycosaminoglycans from kidney cortex from healthy db/+ and diabetic
Nonheparin anticoagulants for heparin-induced thrombocytopenia.
John G Kelton et al.
The New England journal of medicine, 368(8), 737-744 (2013-02-22)
Xue Zhao et al.
The Journal of biological chemistry, 288(13), 9226-9237 (2013-02-21)
Decorin proteoglycan is comprised of a core protein containing a single O-linked dermatan sulfate/chondroitin sulfate glycosaminoglycan (GAG) chain. Although the sequence of the decorin core protein is determined by the gene encoding its structure, the structure of its GAG chain
A Bajetto et al.
Journal of biological regulators and homeostatic agents, 27(1), 143-154 (2013-03-16)
Glioblastoma multiforme (GBM) is among the most devastating human tumors being rapidly fatal despite aggressive surgery, radiation and chemotherapies. It is characterized by extensive dissemination of tumor cells within the brain that hinders complete surgical resection. GBM tumor initiating-cells (TICs)
Martin A Thelin et al.
The FEBS journal, 280(10), 2431-2446 (2013-02-28)
The presence of iduronic acid in chondroitin/dermatan sulfate changes the properties of the polysaccharides because it generates a more flexible chain with increased binding potentials. Iduronic acid in chondroitin/dermatan sulfate influences multiple cellular properties, such as migration, proliferation, differentiation, angiogenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service