Skip to Content
Merck
All Photos(1)

Documents

Y0000586

Valnemulin impurity E

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Pleuromutilin, Drosophilin B, Mutilin 14-glycolate, NSC 121145

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O5
CAS Number:
Molecular Weight:
378.50
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

valnemulin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1CC[C@@]23CCC(=O)[C@H]2[C@]1(C)[C@@H](C[C@@](C)(C=C)[C@@H](O)[C@@H]3C)OC(=O)CO

InChI

1S/C22H34O5/c1-6-20(4)11-16(27-17(25)12-23)21(5)13(2)7-9-22(14(3)19(20)26)10-8-15(24)18(21)22/h6,13-14,16,18-19,23,26H,1,7-12H2,2-5H3/t13-,14+,16-,18+,19+,20-,21+,22+/m1/s1

InChI key

ZRZNJUXESFHSIO-BKUNHTPHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Valnemulin impurity E EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Pleuromutilin is an antibiotic natural product that inhibits bacterial protein synthesis by binding to bacterial ribosomes in the peptidyl transferase center and inhibiting peptide bond formation.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruofeng Shang et al.
Journal of medicinal chemistry, 57(13), 5664-5678 (2014-06-04)
A series of novel pleuromutilin derivatives possessing thiadiazole moieties were synthesized via acylation reactions under mild conditions. The in vitro antibacterial activities of the derivatives against methicillin-resistant Staphylococcus aureus, methicillin-resistant Staphylococcus epidermidis, Escherichia coli, and Streptococcus agalactiae were tested by
Rodger Novak et al.
Current opinion in investigational drugs (London, England : 2000), 11(2), 182-191 (2010-01-30)
Pleuromutilins were discovered as natural-product antibiotics in 1950. Tiamulin was the first pleuromutilin compound to be approved for veterinary use in 1979, followed by valnemulin in 1999. It was not until 2007 that retapamulin became the first pleuromutilin approved for
Rodger Novak
Annals of the New York Academy of Sciences, 1241, 71-81 (2011-12-24)
In 1951, the first reference to the antibacterial substance pleuromutilin was made in a paper published in the Proceedings of the National Academy of Sciences. Researchers had identified several species of the mold genus Pleurotus that inhibited the growth of
Y-Z Tang et al.
Mini reviews in medicinal chemistry, 12(1), 53-61 (2011-11-11)
Due to the rapid onset of resistance to most antibacterial drugs, research efforts are focusing on new classes of antibacterials with different mechanisms of action from clinically used antibacterials. Pleuromutilin derivatives have received more and more scientific attention for their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service