Skip to Content
Merck
All Photos(1)

Key Documents

Y0000485

myo-Inositol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

myo-Inositol, 1,2,3,4,5,6-Hexahydroxycyclohexane, i-Inositol, meso-Inositol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1907329
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

vapor density

6.2 (vs air)

API family

inositol

manufacturer/tradename

EDQM

mp

222-227 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O[C@H]1[C@H](O)[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4+,5-,6-

InChI key

CDAISMWEOUEBRE-GPIVLXJGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

myo-Inositol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

A component of membrane phospholipids, glycosyl­phosphatidyl­inositol anchors that bind glycoproteins to cell membranes, and inositol phosphate second messengers.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksey Porollo et al.
mBio, 5(6), e01834-e01834 (2014-11-06)
In the context of deciphering the metabolic strategies of the obligate pathogenic fungi in the genus Pneumocystis, the genomes of three species (P. carinii, P. murina, and P. jirovecii) were compared among themselves and with the free-living, phylogenetically related fission
Ethan S Patterson et al.
Developmental biology, 393(1), 44-56 (2014-07-06)
Geminin is a nucleoprotein that can directly bind chromatin regulatory complexes to modulate gene expression during development. Geminin knockout mouse embryos are preimplantation lethal by the 32-cell stage, precluding in vivo study of Geminin's role in neural development. Therefore, here
Alan Winston et al.
PloS one, 10(2), e0118608-e0118608 (2015-02-28)
Changes in cerebral metabolite ratios (CMR) measured on 1H-MRS and changes in cognitive function (CF) are described in subjects commencing combination antiretroviral therapy (cART), although the dynamics of such changes are poorly understood. Neuroasymptomatic, HIV-infected subjects electively commencing cART were
Ming Huang et al.
Journal of plant physiology, 171(14), 1205-1216 (2014-07-16)
Chestnut rose (Rosa roxburghii Tratt) is a fruit crop that contains unusually high levels of l-ascorbic acid (AsA; ∼1300 mg 100g(-1) FW). To explore the mechanisms underlying AsA metabolism, we investigated the distribution and abundance of AsA during fruit development.
Jennifer J Donegan et al.
Molecular pharmacology, 87(3), 492-500 (2015-01-01)
The serotonin 2A (5-HT2A) receptor and the proinflammatory cytokine, interleukin-6 (IL-6), have both been implicated in psychiatric disorders. Previously, we demonstrated that these molecules both facilitate cognitive flexibility, a prefrontal cortex-mediated executive function impaired in multiple mental illnesses. In this

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service