Skip to Content
Merck
All Photos(1)

Key Documents

Y0000415

Felypressin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-L-Phenylalanine-8-L-lysinevasopressin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H65N13O11S2
CAS Number:
Molecular Weight:
1040.22
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

felypressin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

S1SCC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C1)C(=O)N4C(CCC4)C(=O)NC(CCCCN)C(=O)NCC(=O)N)CC(=O)N)CCC(=O)N)Cc3ccccc3)Cc2ccccc2)N

InChI

1S/C46H65N13O11S2/c47-18-8-7-14-29(40(64)52-23-38(51)62)54-45(69)35-15-9-19-59(35)46(70)34-25-72-71-24-28(48)39(63)55-31(20-26-10-3-1-4-11-26)43(67)56-32(21-27-12-5-2-6-13-27)42(66)53-30(16-17-36(49)60)41(65)57-33(22-37(50)61)44(68)58-34/h1-6,10-13,28-35H,7-9,14-25,47-48H2,(H2,49,60)(H2,50,61)(H2,51,62)(H,52,64)(H,53,66)(H,54,69)(H,55,63)(H,56,67)(H,57,65)(H,58,68)

InChI key

SFKQVVDKFKYTNA-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Felypressin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camila A Fleury et al.
Arquivos brasileiros de cardiologia, 114(2), 295-303 (2020-03-28)
Cigarette smoking is usually associated with hypertension and may modify vasoconstrictor response. The present study aimed to analyze and compare the interaction of passive cigarette smoking and hypertension on epinephrine and felypressin blood pressure effects after intravascular injection. 45-day male
Wai Yoong et al.
Journal of minimally invasive gynecology, 24(5), 811-814 (2017-04-16)
To examine demographics and outcome measures of women having undergone vaginal excision of myomas through the Dührssen (longitudinal median cervical) incision. Prospective case series (Canadian Task Force classification II-3). A London teaching hospital. Nineteen patients with either a submucous myoma
N S Oliveira et al.
Pharmacological research, 48(4), 325-328 (2003-08-07)
This study aimed to observe the effects of prilocaine chloridate (P) and the associations of P with felypressin (P+F), and epinephrine (P+E) on the heart rate and force of contraction on the isolated hearts of rats. The hearts were perfused
N S Oliveira et al.
Pharmacological research, 46(1), 15-18 (2002-09-05)
Some reports have demonstrated that the vasoconstrictor effect of catecholamines may be amplified by felypressin. Thus, the association of felypressin and epinephrine with local anesthetics may be indicated to achieve more effective vasoconstrictor actions with fewer collateral effects from both
Javier Caviedes-Bucheli et al.
Journal of endodontics, 35(5), 631-633 (2009-05-05)
The purpose of this study was to quantify the effect of the infiltration injection of different vasoconstrictor and anesthetic solutions on substance P (SP) expression in healthy human dental pulp. Thirty pulp samples were obtained from healthy upper premolars in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service