Skip to Content
Merck
All Photos(1)

Key Documents

Y0000412

Sulfamethoxazole impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N4-Acetylsulfamethoxazole, N-Acetyl sulfamethoxazole, Acetyl-sulfamethoxazole, N-{4-{[(5-Methyl-3-isoxazolyl)amino]sulfonyl}phenyl}acetamide, Sulfisomezole-N4-acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N3O4S
CAS Number:
Molecular Weight:
295.31
Beilstein:
285801
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulfamethoxazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=S(NC1=NOC(C)=C1)(C2=CC=C(NC(C)=O)C=C2)=O

InChI

1S/C12H13N3O4S/c1-8-7-12(14-19-8)15-20(17,18)11-5-3-10(4-6-11)13-9(2)16/h3-7H,1-2H3,(H,13,16)(H,14,15)

InChI key

GXPIUNZCALHVBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfamethoxazole impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T B Vree et al.
The Veterinary quarterly, 9(4), 381-384 (1987-10-01)
The turtle Pseudemys scripta elegans is able to hydroxylate and acetylate Sulphamethoxazole in a way that is comparable to man, i.e. the rate and yield of hydroxylation equals that of the acetylation. The hydroxy metabolites 5-hydroxy- and N4-acetyl-5-hydroxysulphamethoxazole are not
O Spreux-Varoquaux et al.
Journal of chromatography, 274, 187-199 (1983-05-13)
A normal-phase high-performance liquid chromatographic method was developed to determine therapeutic concentrations of trimethoprim, sulphamethoxazole, and its N4-acetyl derivative in biological fluids. The compounds are extracted at pH 6.2 using ethyl acetate--chloroform in a single extraction. The detection limit is
Ion-paired high-performance liquid chromatographic separation of trimethoprim, sulfamethoxazole and N4-acetylsulfamethoxazole with solid-phase extraction.
S C Laizure et al.
Journal of chromatography, 528(1), 235-242 (1990-06-08)
J Blaser et al.
Infection, 21(4), 206-209 (1993-07-01)
Serum kinetics of trimethoprim and sulfamethoxazole were studied in 23 patients during oral and i.v. treatment of Pneumocystis carinii pneumonia. Daily doses of 15-22 mg/kg trimethoprim and 75-110 mg/kg sulfamethoxazole were given every 6 h. Despite administration of a loading
R M Hutabarat et al.
Clinical pharmacology and therapeutics, 49(4), 402-409 (1991-04-01)
The disposition of sulfamethoxazole and trimethoprim, after constant rate intravenous administration (10 mg/kg/hr sulfamethoxazole and 2 mg/kg/hr trimethoprim for 1 hour), was investigated in adult patients with cystic fibrosis (n = 7) and in age-matched healthy subjects (control subjects, n

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service