Skip to Content
Merck
All Photos(1)

Key Documents

Y0000320

Clarithromycin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H69NO13
CAS Number:
Molecular Weight:
747.95
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clarithromycin

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

SMILES string

O[C@@H]([C@@](O)(C)[C@@H](CC)OC([C@H](C)[C@H]1O[C@@H]2O[C@@H](C)[C@H](O)[C@](C)(OC)C2)=O)[C@@H](C)C([C@H](C)C[C@@](C)(OC)[C@H](O[C@@H]3O[C@H](C)C[C@H](N(C)C)[C@H]3O)[C@H]1C)=O

InChI

1S/C38H69NO13/c1-15-26-38(10,45)31(42)21(4)28(40)19(2)17-37(9,47-14)33(52-35-29(41)25(39(11)12)16-20(3)48-35)22(5)30(23(6)34(44)50-26)51-27-18-36(8,46-13)32(43)24(7)49-27/h19-27,29-33,35,41-43,45H,15-18H2,1-14H3/t19-,20-,21+,22+,23-,24+,25+,26-,27+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1

InChI key

AGOYDEPGAOXOCK-KCBOHYOISA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clarithromycin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Clarithromycin is a macrolide antibiotic that interferes with bacterial protein synthesis. Clarithromycin is an acid-stable version of erythromycin and is particularly effective against Gram-negative bacteria. It has a short half-life, however its metabolite, 14-hydroxy clarithromycin is nearly twice as active as clarithromycin against certain bacteria.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleonora Altman et al.
Canadian journal of microbiology, 60(1), 35-39 (2014-01-08)
Lipopolysaccharide (LPS) of Helicobacter pylori exhibits several unique structures, such as Lewis (Le) antigens, α-1,6-glucan, and dd-heptan. To investigate the relationship between LPS structure and resistance to clarithromycin, 41 Canadian isolates of H. pylori were characterized by whole-cell ELISA (enzyme-linked
Stig Lyngbæk et al.
European journal of preventive cardiology, 21(10), 1275-1284 (2013-06-01)
Low prevalence of detectable cardiac troponin in healthy people and low-risk patients previously curtailed its use. With a new high-sensitive cardiac troponin assay (hs-cTnT), concentrations below conventional detection may have prognostic value, notably in combination with N-terminal pro-B-type natriuretic peptide
Shamira J Shallom et al.
Journal of clinical microbiology, 51(9), 2943-2949 (2013-06-28)
Mycobacterium abscessus (M. abscessus sensu lato, or the M. abscessus group) comprises three closely related taxa whose taxonomic statuses are under revision, i.e., M. abscessus sensu stricto, Mycobacterium bolletii, and Mycobacterium massiliense. We describe here a simple, robust, and cost-effective
Chung-Su Park et al.
The American journal of gastroenterology, 109(10), 1595-1602 (2014-08-06)
Helicobacter pylori eradication rates with clarithromycin-based triple therapy are declining, and an alternative strategy is needed urgently. We sought to compare the efficacy of pretreatment antimicrobial susceptibility-guided vs. clarithromycin-based triple therapy for H. pylori eradication in a region with high
Kentaro Nagaoka et al.
Antimicrobial agents and chemotherapy, 57(4), 1844-1849 (2013-02-06)
Fusobacterium nucleatum is one of the most common anaerobic bacteria in periodontitis and is responsible for several extraoral infections, including respiratory tract diseases. In this study, we examined whether F. nucleatum induces mucin secretion in airway epithelial cells. We also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service