Skip to Content
Merck
All Photos(1)

Key Documents

Y0000279

Tri-n-butyl phosphate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tributyl phosphate, TBP, TBPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
Beilstein:
1710584
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

vapor density

9.2 (vs air)

vapor pressure

27 mmHg ( 178 °C)
7.3 mmHg ( 150 °C)

API family

tributyl phosphate

autoignition temp.

770 °F

manufacturer/tradename

EDQM

refractive index

n20/D 1.424 (lit.)

bp

180-183 °C/22 mmHg (lit.)

mp

−79 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCOP(=O)(OCCCC)OCCCC

InChI

1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3

InChI key

STCOOQWBFONSKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tri-n-butyl phosphate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Burnouf et al.
Vox sanguinis, 104(1), 1-6 (2012-07-05)
A minipool solvent/detergent (S/D; 1% TnBP/1% Triton X-45; 31°C) process was developed for viral inactivation of plasma and cryoprecipitate used for transfusion. The goal of this study was to determine the rate and extent of inactivation of dengue virus (DENV)
Domenico Santoro et al.
Environmental science & technology, 44(16), 6233-6241 (2010-08-14)
Advanced Oxidation Processes (AOPs) promoted by ultraviolet light are innovative and potentially cost-effective solutions for treating persistent pollutants recalcitrant to conventional water and wastewater treatment. While several studies have been performed during the past decade to improve the fundamental understanding
C W Van Neste et al.
Analytical chemistry, 81(5), 1952-1956 (2009-02-04)
Despite its immediate applications, selective detection of trace quantities of surface adsorbed chemicals, such as explosives, without physically collecting the sample molecules is a challenging task. Standoff spectroscopic techniques offer an ideal method of detecting chemicals without using a sample
Muniasamy Neerathilingam et al.
Toxicology letters, 199(1), 10-16 (2010-08-07)
Tributyl phosphate (TBP) is a toxic organophosphorous compound widely used in many industrial applications, including significant usage in nuclear processing. The industrial application of this chemical is responsible for occupational exposure and environmental pollution. In this study, (1)H NMR-based metabonomics
Torsten Klengel et al.
Nature neuroscience, 16(1), 33-41 (2012-12-04)
Although the fact that genetic predisposition and environmental exposures interact to shape development and function of the human brain and, ultimately, the risk of psychiatric disorders has drawn wide interest, the corresponding molecular mechanisms have not yet been elucidated. We

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service