Skip to Content
Merck
All Photos(1)

Key Documents

Y0000257

Lactobionic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-O-β-D-Galactopyranosyl-D-gluconic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O12
CAS Number:
Molecular Weight:
358.30
Beilstein:
95054
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lactobionic acid

manufacturer/tradename

EDQM

mp

113-118 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C(O)=O

InChI

1S/C12H22O12/c13-1-3(15)10(7(18)8(19)11(21)22)24-12-9(20)6(17)5(16)4(2-14)23-12/h3-10,12-20H,1-2H2,(H,21,22)/t3-,4-,5+,6+,7-,8-,9-,10-,12+/m1/s1

InChI key

JYTUSYBCFIZPBE-AMTLMPIISA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. For further information and support, including product information leaflets, please go to the website of the issuing Pharmacopoeia.

Application

Lactobionic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberley A Stannard et al.
Cancer letters, 299(2), 95-110 (2010-09-10)
High level galectin-1 expression results in cancer cell evasion of the immune response, increased tumour survival and aggressive metastases. Using a galectin-1 polyclonal antibody, high levels of galectin-1 protein were shown to be expressed by breast cancer cells established from
K M Kamruzzaman Selim et al.
Journal of materials science. Materials in medicine, 20(9), 1945-1953 (2009-04-15)
In the current study, beta-galactose-carrying lactobionic acid (LA) was conjugated on the surface of mercaptoacetic acid-coated cadmium sulfide nanoparticles (CSNPs) to ensure specific recognition of liver cells (hepatocytes) and to enhance biocompatibility. Maltotrionic acid-coated CSNPs (MCSNPs) were also prepared for
Xinru Li et al.
Journal of drug targeting, 17(10), 739-750 (2009-10-30)
Polymeric micelles, based on lactobionic acid (LA)-conjugated Pluronic P105 (P105), were prepared to achieve liver-targeted delivery of silybin. In the triblock copolymer structure of PEO-PPO-PEO, LA was successfully conjugated with the terminal end of PEO to produce LA-P105. The success
Israel Pedruzzi et al.
Enzyme and microbial technology, 49(2), 183-191 (2011-11-25)
In this work, we have investigated the kinetics of the biotechnological production of lactobionic acid (LBA) and sorbitol by the catalytic action of glucose-fructose oxidoreductase (GFOR) and glucono-δ-lactonase (GL) enzymes. The cells of bacterium Zymomonas mobilis ATCC 29191 containing this
Saúl Alonso et al.
Biotechnology advances, 31(8), 1275-1291 (2013-05-09)
Lactobionic acid has appeared on the commercial scene as a versatile polyhydroxy acid with numerous promising applications in the food, medicine, pharmaceutical, cosmetics and chemical industries. This high value-added bio-product has recently received growing attention as a bioactive compound, providing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service