Skip to Content
Merck
All Photos(1)

Key Documents

Y0000149

Rifabutin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ansamycin, Ansatipine (Farmitalia), LM-427, Mycobutin (Farmitalia)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H62N4O11
CAS Number:
Molecular Weight:
847.00
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

rifabutin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[N+H]1(CCC2(NC3=C4NC(=O)\C(=C/C=C/[C@@H]([C@@H]([C@H]([C@H]([C@H]([C@@H]([C@@H]([C@H](\C=C\O[C@]5(Oc6c(c(c(c(c6C)[O-])C4=O)C3=N2)C5=O)C)OC)C)OC(=O)C)C)O)C)O)C)\C)CC1)CC(C)C

InChI

1S/C46H62N4O11/c1-22(2)21-50-18-16-46(17-19-50)48-34-31-32-39(54)28(8)42-33(31)43(56)45(10,61-42)59-20-15-30(58-11)25(5)41(60-29(9)51)27(7)38(53)26(6)37(52)23(3)13-12-14-24(4)44(57)47-36(40(32)55)35(34)49-46/h12-15,20,22-23,25-27,30,37-38,41,49,52-54H,16-19,21H2,1-11H3,(H,47,57)/b13-12+,20-15+,24-14-/t23-,25+,26+,27+,30-,37-,38+,41+,45-/m0/s1

InChI key

ATEBXHFBFRCZMA-VXTBVIBXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rifabutin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Rifabutin is an antibiotic; antitumor. Rifabutin interferes with HSP-90 molecular chaperone, enhances ubiquitination and protein degradation, and inactivates bacterial RNA polymerase.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Dautzenberg
Infection, 25(1), 63-66 (1997-01-01)
The MICs of rifabutin on Mycobacterium avium are compatible with its efficacy in clinical infections. Two North American trials established the prophylactic effect of rifabutin in disseminated M. avium disease in AIDS patients. Several prospective non-randomized trials show the clinical
R E Chaisson
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 22 Suppl 1, S61-S66 (1996-04-01)
Patients with advanced human immunodeficiency virus (HIV) disease are at risk for infections caused by protozoa, fungi, viruses, bacteria, and mycobacteria. Chemoprophylaxis is being used increasingly to prevent a growing number of opportunistic infections that occur in HIV-infected patients. Multiple
Anne M Baciewicz et al.
The American journal of the medical sciences, 335(2), 126-136 (2008-02-16)
Rifampin is a potent inducer of cytochrome P-450 oxidative enzymes as well as the P-glycoprotein transport system. Several examples of well-documented clinically significant interactions include warfarin, oral contraceptives, cyclosporine, itraconazole, digoxin, verapamil, nifedipine, simvastatin, midazolam, and human immunodeficiency virus-related protease
Is rifabutin prophylaxis against Mycobacterium avium complex infection in HIV infection worthwhile? The net impact on patients suggests not.
J M Stephenson et al.
Genitourinary medicine, 72(4), 272-276 (1996-08-01)
G Davies et al.
The Cochrane database of systematic reviews, (4)(4), CD005159-CD005159 (2007-10-19)
Rifamycins are an essential component of modern short-course regimens for treating tuberculosis. Rifabutin has favourable pharmacokinetic and pharmacodynamic properties and is less prone to drug-drug interactions than rifampicin. It could contribute to shortening of therapy or simplify treatment in HIV-positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service