Skip to Content
Merck
All Photos(1)

Key Documents

Y0000141

Ioxaglic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-(2-Hydroxyethyl)-2,4,6-triiodo-5-{2-[2,4,6-triiodo-3-(N-methylacetamido)-5-(methylcarbamoyl)benzamido]acetamido}isophthalamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21I6N5O8
CAS Number:
Molecular Weight:
1268.88
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ioxaglic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Ic1c(c(c(c(c1C(=O)NCC(=O)Nc2c(c(c(c(c2I)C(=O)O)I)C(=O)NCCO)I)I)C(=O)NC)I)N(C)C(=O)C

InChI

1S/C24H21I6N5O8/c1-7(37)35(3)20-17(29)10(21(39)31-2)13(25)11(18(20)30)23(41)33-6-8(38)34-19-15(27)9(22(40)32-4-5-36)14(26)12(16(19)28)24(42)43/h36H,4-6H2,1-3H3,(H,31,39)(H,32,40)(H,33,41)(H,34,38)(H,42,43)

InChI key

TYYBFXNZMFNZJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ioxaglic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Kerckhofs et al.
European cells & materials, 25, 179-189 (2013-02-08)
We describe a non-destructive imaging method, named contrast-enhanced nanofocus X-ray computed tomography (CE-nanoCT), that permits simultaneously imaging and quantifying in 3D the (sub)tissue architecture and (biochemical) composition of cartilage and bone in small animal models at a novel contrast and
Laura Calvi et al.
Thyroid : official journal of the American Thyroid Association, 21(4), 443-449 (2011-03-10)
Thyrotoxicosis caused by destructive thyroiditis is self-limited and results from the subacute release of preformed thyroid hormone. Common etiologies include painful subacute thyroiditis and silent (painless) subacute thyroiditis (including postpartum thyroiditis, amiodarone-associated destructive thyroiditis, and lithium-associated thyroiditis). Thyrotoxicosis commonly evolves
Christian Delcour et al.
Cardiovascular and interventional radiology, 36(1), 237-238 (2012-06-15)
To test the compatibility of trisodium citrate, a catheter lock solution, with iodinated contrast medium. Iohexol, iobitridol, iodixanol, ioxaglate, ioxithalamate, iomeprol, and iopromide were tested. In all tests, 2 ml of contrast medium were mixed with 2 ml of trisodium citrate solution.
M Siebelt et al.
Osteoarthritis and cartilage, 19(10), 1183-1189 (2011-08-09)
Similar to delayed gadolinium enhanced MRI of cartilage, it might be possible to image cartilage quality using CT arthrography (CTa). This study assessed the potential of CTa as a clinically applicable tool to evaluate cartilage quality in terms of sulphated
Emre Aslanger et al.
Coronary artery disease, 23(4), 265-270 (2012-02-22)
Contrast medium-induced nephropathy (CIN) is a well-known complication of coronary angiographic procedures, especially in patients treated with primary angioplasty. To prevent CIN, we examined using a local application of N-acetylcysteine (NAC) for the prevention of CIN during primary angioplasty. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service