Skip to Content
Merck
All Photos(1)

Key Documents

Y0000049

Amlodipine besylate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-[(2-Aminoethoxy)-methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5- pyridinedicarboxylic acid 3-ethyl 5-methyl ester benzene sulfonate, Norvasc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25ClN2O5 · C6H5SO3H
CAS Number:
Molecular Weight:
567.05
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

amlodipine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OS(=O)(=O)c1ccccc1.CCOC(=O)C2=C(COCCN)NC(C)=C(C2c3ccccc3Cl)C(=O)OC

InChI

1S/C20H25ClN2O5.C6H6O3S/c1-4-28-20(25)18-15(11-27-10-9-22)23-12(2)16(19(24)26-3)17(18)13-7-5-6-8-14(13)21;7-10(8,9)6-4-2-1-3-5-6/h5-8,17,23H,4,9-11,22H2,1-3H3;1-5H,(H,7,8,9)

InChI key

ZPBWCRDSRKPIDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amlodipine besylate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Amlodipine is an L-type calcium channel blocker. Amlodipine belongs to a class of cardiovascular drugs, which act at the voltage gated calcium channel of the CaV1, or L-type, class. Amlodipine also has antihypertensive and antianginal effects. Its activity resides mainly in the (-)-isomer. Amlodipine inhibits growth of human epidermoid carcinoma A431 cells and has antireproductive effects in male rats.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Cooper et al.
The New England journal of medicine, 370(1), 13-22 (2013-11-20)
Atherosclerotic renal-artery stenosis is a common problem in the elderly. Despite two randomized trials that did not show a benefit of renal-artery stenting with respect to kidney function, the usefulness of stenting for the prevention of major adverse renal and
László Bajnok
Orvosi hetilap, 154(7), 243-247 (2013-02-12)
From the evaluated ONTARGET, ALTITUDE, ACCOMPLISH, ROADMAP, and ACCORD-BP studies a conclusion can be drawn that though microalbuminuria/proteinuria is a strong epidemiological biomarker, in interventional studies it is not necessarily a reliable surrogate endpoint as actual renal function may change
Juliano Lara Fernandes et al.
The American journal of medicine, 126(9), 834-837 (2013-07-09)
Iron chelation therapy in patients with thalassemia major may not prevent iron overload in all organs, especially those in which iron enters cells through specific calcium channels. We designed a controlled pilot study to assess the potential of the calcium
S Neldam et al.
International journal of clinical practice, 67(9), 843-852 (2013-08-21)
Rapid and sustained blood pressure (BP) goal attainment is important to reduce cardiovascular risk. Initial use of combination therapy may improve BP goal attainment. The Boehringer Ingelheim trial database was searched for randomised, double-blind studies comparing telmisartan/amlodipine combination therapy with
Giuseppe Derosa et al.
Journal of the American Society of Hypertension : JASH, 7(1), 32-39 (2013-01-17)
The purpose of this study was to evaluate a fixed olmesartan/amlodipine combination on blood pressure control, lipid profile, insulin sensitivity, and some inflammatory markers compared with single-drug monotherapy. A total of 276 hypertensive patients were randomly assigned to olmesartan 20

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service