Skip to Content
Merck
All Photos(1)

Key Documents

X100PC

Sigma-Aldrich

Triton X-100

peroxide- and carbonyl-free

Synonym(s):

t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

description

non-ionic

Quality Level

form

liquid

mol wt

micellar avg mol wt 80,000
average mol wt 625

packaging

ampule of 5, 5x5, 10x5 mL

aggregation number

100-155

CMC

0.2-0.9 mM (20-25°C)

transition temp

cloud point 65 °C
pour point ~7 °C

solubility

water: soluble

HLB

13.5

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triton X-100 is a common non-ionic surfactant and emulsifier which is often used in biochemical applications to solubilize proteins. It is considered a comparatively mild detergent, non-denaturing, and is reported in numerous references as a routinely added reagent. It is utilized for lysing cells to extract protein and cellular organelles. It can also permeabilize the living cell membrane for transfection.[1][2]

Application

0·3% Triton X-100 in TBS has been used for permeabilization of retinas before immunostaining.[3][4] 0.5% Triton X-100 has been used for astrocyte cell lysis.[5]

Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Packaging

Packaged in ampules under Argon.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Quantitative ex vivo detection of rodent retinal ganglion cells by immunolabeling Brn-3b.
Experimental Eye Research, 79(1), 131-140 (2004)
Cytotoxicity of bilirubin for human fibroblasts and rat astrocytes in culture. Effect of the ratio of bilirubin to serum albumin.
Chuniaud L, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 256(2), 103-114 (1996)
Adeno-associated virus mediated SOD gene therapy protects the retinal ganglion cells from chronic intraocular pressure elevation induced injury via attenuating oxidative stress and improving mitochondrial dysfunction in a rat model.
American Journal of Translational Research, 8(2), 799-799 (2016)
The size, shape, and hydration of nonionic surfactant micelles. Triton X-100.
Robson RJ and Dennis EA
The Journal of Physical Chemistry, 81, 1075-1078 (1977)
Friederike Pfeiffer et al.
Acta neuropathologica, 122(5), 601-614 (2011-10-11)
In experimental autoimmune encephalomyelitis (EAE), an animal model for multiple sclerosis (MS), loss of the blood-brain barrier (BBB) tight junction (TJ) protein claudin-3 correlates with immune cell infiltration into the CNS and BBB leakiness. Here we show that sealing BBB

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service