Skip to Content
Merck
All Photos(1)

Key Documents

W237712

Sigma-Aldrich

Diethyl succinate

natural, ≥99%, FG

Synonym(s):

Diethyl butanedioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH2CH2COOC2H5
CAS Number:
Molecular Weight:
174.19
FEMA Number:
2377
Beilstein:
907645
EC Number:
Council of Europe no.:
438
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.444
NACRES:
NA.21

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

6 (vs air)

Assay

≥99%

refractive index

n20/D 1.42 (lit.)

bp

218 °C (lit.)

mp

−20 °C (lit.)

density

1.047 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; grape; waxy; fruity; floral

SMILES string

CCOC(=O)CCC(=O)OCC

InChI

1S/C8H14O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-6H2,1-2H3

InChI key

DKMROQRQHGEIOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl succinate (DES) is a diester. This chemical can be used as a food additive, fragrance, and flavoring agent . It is a key component of wine aroma. Natural sources of this compound include apples, grapes, and cocoa .

Application

Diethyl succinate:
  • Contribute to the aroma of sparkling wine.
  • is a key flavor component of rice wine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of the changes in volatile compounds, aroma and sensory attributes during the production process of sparkling wine by traditional method
Ubeda C, et al.
Food Research International, 119, 554-563 (2019)
Effect of mixed yeast starter on volatile flavor compounds in Chinese rice wine during different brewing stages
Yang Y, et al.
LWT--Food Science and Technology, 78, 373-381 (2017)
Phase equilibrium measurements and thermodynamic modeling of {CO2 + diethyl succinate + cosolvent} systems
loc AP, et al.
Fluid Phase Equilibria, 502, 112285-112285 (2019)
Effects of different techniques of malolactic fermentation induction on diacetyl metabolism and biosynthesis of selected aromatic esters in cool-climate grape wines
Lasik-Kurdys, M, et al.
Molecules (Basel), 23(10), 2549-2549 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service