Skip to Content
Merck
All Photos(1)

Key Documents

W1761

Millipore

Wilkins Chalgren anaerobic agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85
Pricing and availability is not currently available.

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.1±0.2 (25 °C)

application(s)

cleaning products
clinical testing
cosmetics
food and beverages
personal care

microbiology

suitability

anaerobic bacteria
selective for Bacteroides spp.
selective for Bifidobacterium spp.
selective for Clostridium spp.

Application

Used for the isolation of anaerobic bacteria.

Components

Ingredients (g/L)
Casein enzymatic hydrolysate, 10.00
Peptic digest of animal tissue, 10.00
Yeast extract, 5.00
Dextrose, 1.00
Sodium chloride, 5.00
L-Arginine, 1.00
Sodium pyruvate, 1.00
Hemin, 0.005
Menadione, 0.0005
Agar, 10.00

Preparation Note

Suspend 43 g of Wilkins Chalgren Anaerobic Agar in 1000 ml of distilled water. Heat to boiling to dissolve the medium completely. Dispense and sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. Cool to 50°C before adding antibiotics to be tested. Mix gently and pour into sterile petri plates.For the cultivation of anaerobes add 2 vial of Non-Spore Anaerobic Supplement (Cat. No. N8781) or 2 vial of G.N. Spore Anaerobic Supplement (Cat. No. G3789) as desired into the sterile molten medium before pouring into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T D Wilkins et al.
Antimicrobial agents and chemotherapy, 10(6), 926-928 (1976-12-01)
A medium is described which was designed for use in testing the minimal inhibitory concentration of antibiotics for anaerobic bacteria by agar dilution. It contains: Trypticase (1%), Gelysate (1%), yeast extract (0.5%), glucose (0.1%), pyruvate (0.1%), arginine (0.1%), NaCl (0.5%)

Articles

Bifidobacteria-selective media detects beneficial bacteria, aiding quality control in microbiology labs.

Bifidobacteria-selective media detects beneficial bacteria, aiding quality control in microbiology labs.

Bifidobacteria-selective media detects beneficial bacteria, aiding quality control in microbiology labs.

Bifidobacteria-selective media detects beneficial bacteria, aiding quality control in microbiology labs.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service