UPMGCL2
Ultra pure 25mM MgCl2
DNA-free, Nuclease-free 25mM MgCl2, designed for further manufacture
About This Item
Recommended Products
Quality Level
form
liquid
feature
DNA free
RNase and DNase free (also Nickase free)
packaging
pkg of 1.5 mL
technique(s)
RT-PCR: suitable
cDNA synthesis: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Ultra pure 25mM MgCl2 is manufactured under stringent conditions to ensure that it is free of detectable levels of DNA, DNase, RNase, and Nickase to reduce the risk of false positive and false negative results in molecular diagnostic assays. When used with Ultra pure 10x Taq Buffer without MgCl2, Ultra pure 25mM MgCl2 allows flexibility to optimize amplification conditions through adjustments to MgCl2 concentration. Ultra pure 25mM MgCl2 should be used with other ultra pure DNA-free reagents, such as Ultra pure JumpStart™ Taq DNA Polymerase and Ultra pure DNA-free Water, to minimize false positive and negative results and improved DNA amplification.
Ultra pure 25mM MgCl2 was designed for use in highly-regulated industries in applications that require a high level of performance, lot-to-lot consistency, and high sensitivity without false results from reagent-borne contamination while meeting industry-driven critical regulations. To meet this requirements we have increased the quality level of our products. The M-Clarity Program defines product quality. It provides transparency so that you can choose, with confidence, suitable products that meet your needs with respect to quality and regulatory compliance, transparency, change notification, and documentation.
Application
- Quantitative PCR
- Amplification of complex genomic DNA and cDNA templates
- Amplification of low-copy number targets
- Microbiome analysis
- For further manufacture
Features and Benefits
- DNA-free
- DNase-free, RNase-free, and nickase-free
- Ideal for high throughput quantitative PCR applications
- Compatible with Ultra pure JumpStart™ Taq DNA Polymerase
- Flexibility to optimize amplification conditions
- Enhanced change control and quality agreement
Packaging
Other Notes
Legal Information
related product
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Utilize molecular raw materials for diagnostic assay development and manufacturing, backed by extensive experience and worldwide manufacturing.
Utilize molecular raw materials for diagnostic assay development and manufacturing, backed by extensive experience and worldwide manufacturing.
Utilize molecular raw materials for diagnostic assay development and manufacturing, backed by extensive experience and worldwide manufacturing.
Utilize molecular raw materials for diagnostic assay development and manufacturing, backed by extensive experience and worldwide manufacturing.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service