T5509
Trigonelline hydrochloride
analytical standard
Synonym(s):
1-Methylpyridinium-3-carboxylate hydrochloride, N-Methylnicotinic acid betaine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H7NO2 · HCl
CAS Number:
Molecular Weight:
173.60
Beilstein:
3734169
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
food and beverages
format
neat
SMILES string
[Cl-].C[n+]1cccc(c1)C(O)=O
InChI
1S/C7H7NO2.ClH/c1-8-4-2-3-6(5-8)7(9)10;/h2-5H,1H3;1H
InChI key
TZSYLWAXZMNUJB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A Arlt et al.
Oncogene, 32(40), 4825-4835 (2012-10-31)
Evidence accumulates that the transcription factor nuclear factor E2-related factor 2 (Nrf2) has an essential role in cancer development and chemoresistance, thus pointing to its potential as an anticancer target and undermining its suitability in chemoprevention. Through the induction of
Arvindkumar E Ghule et al.
International immunopharmacology, 14(4), 740-748 (2012-10-30)
Oxidative stress and apoptotic cell death in kidney have been suggested as contributing factors in the development and complication of diabetes especially in diabetic nephropathy (DN). This study investigated the effects of trigonelline (TG) on the renal functional, morphological changes
Laura Sánchez-Hernández et al.
Journal of agricultural and food chemistry, 58(13), 7489-7496 (2010-06-17)
A capillary electrophoresis method with UV detection was developed for the first time for the determination of the pyridine betaine trigonelline (N-methylnicotinic acid) in seeds and vegetable oils. Analytical characteristics of the method showed its good performance in terms of
Ulla Mueller et al.
Food & function, 2(5), 265-272 (2011-07-23)
Coffee shows distinct antimicrobial activity against several bacterial genera. The present study investigated molecular mechanisms and active ingredients mediating the antimicrobial effect of coffee. Depending on concentration, roasted, but not raw coffee brew inhibited the growth of Escherichia coli and
Laura Sánchez-Hernández et al.
Electrophoresis, 32(11), 1394-1401 (2011-04-27)
A CE-tandem mass spectrometry (MS²) methodology enabling the simultaneous determination of betaines (glycine betaine, trigonelline, proline betaine and total content of carnitines) in vegetable oils was developed. Betaines were derivatized with butanol previous to their baseline separation in 10 min using
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service