Skip to Content
Merck
All Photos(1)

Key Documents

T1810000

Tranexamic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

trans-4-(Aminomethyl)cyclohexanecarboxylic acid, AMCA, AMCHA, HAKU, TAMCHA, Tranexamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2C6H10CO2H
CAS Number:
Molecular Weight:
157.21
Beilstein:
2207452
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

tranexamic acid

manufacturer/tradename

EDQM

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC[C@H]1CC[C@@H](CC1)C(O)=O

InChI

1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)/t6-,7-

InChI key

GYDJEQRTZSCIOI-LJGSYFOKSA-N

Gene Information

human ... PLG(5340)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tranexamic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Grassin-Delyle et al.
British journal of anaesthesia, 111(6), 916-924 (2013-07-25)
Interest in antifibrinolytic tranexamic acid (TA) has grown since the widespread removal of aprotinin, but its dosing during cardiac surgery is still debated. The objectives of this study were to investigate the population pharmacokinetics (PK) of TA given with either
Case scenario: management of trauma-induced coagulopathy in a severe blunt trauma patient.
Jean-Stephane David et al.
Anesthesiology, 119(1), 191-200 (2013-03-22)
Kristen A Matteson et al.
Obstetrics and gynecology, 121(3), 632-643 (2013-05-03)
To compare the effectiveness of nonsurgical abnormal uterine bleeding treatments for bleeding control, quality of life (QOL), pain, sexual health, patient satisfaction, additional treatments needed, and adverse events. MEDLINE, Cochrane databases, and Clinicaltrials.gov were searched from inception to May 2012.
Chih-Hsiang Chang et al.
Clinical orthopaedics and related research, 472(5), 1552-1557 (2014-01-05)
Systemic tranexamic acid can decrease blood loss and rates of transfusion in patients undergoing total hip arthroplasty (THA). However, the efficacy of topical tranexamic acid in THA has only recently been characterized in a small number of studies. The purpose
K Ker et al.
The British journal of surgery, 100(10), 1271-1279 (2013-07-11)
Tranexamic acid (TXA) reduces blood transfusion in surgery but the extent of the reduction in blood loss and how it relates to the dose of TXA is unclear. A systematic review of randomized trials was performed. Data were extracted on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service