Skip to Content
Merck
All Photos(1)

Key Documents

SMB01384

Sigma-Aldrich

DMIP

≥95% (HPLC)

Synonym(s):

1,6-DMIP, 1,6-Dimethyl-1H-imidazo[4,5-b]pyridin-2-amine, 2-Amino-1,6-dimethylimidazo[4,5-b]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10N4
CAS Number:
Molecular Weight:
162.19
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.21

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

solid

shipped in

2-8°C

storage temp.

2-8°C

SMILES string

[n]1(c2c(nc1N)ncc(c2)C)C

InChI

1S/C8H10N4/c1-5-3-6-7(10-4-5)11-8(9)12(6)2/h3-4H,1-2H3,(H2,9,10,11)

InChI key

NEOGGGHDLGYATP-UHFFFAOYSA-N

General description

DMIP (2-Amino-1,6-dimethylimidazo[4,5-b]pyridine) dietary genotoxic carcinogen formed in heat processed meat and fish via the Maillard reaction.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Pais et al.
Journal of agricultural and food chemistry, 47(3), 1098-1108 (1999-12-20)
Mixtures of amino acids, creatine, and glucose simulating the composition of six different kinds of meats (beef, chicken breast, chicken thigh, turkey breast, pork, and fish) were dry-heated to simulate the formation of heterocyclic amines in meats. The presence of
Medjda Bellamri et al.
Genes and environment : the official journal of the Japanese Environmental Mutagen Society, 43(1), 29-29 (2021-07-18)
Heterocyclic aromatic amines (HAAs) form during the high-temperature cooking of meats, poultry, and fish. Some HAAs also arise during the combustion of tobacco. HAAs are multisite carcinogens in rodents, inducing cancer of the liver, gastrointestinal tract, pancreas, mammary, and prostate
Chaoyi Xue et al.
Food research international (Ottawa, Ont.), 155, 111006-111006 (2022-04-12)
The release profiles of protein-bound heterocyclic amines (HAs) in beef myofibril protein (MP) model systems were investigated after different enzyme dosage or stages of digestion were simulated in vitro. The effects of co-digestion of pepper, apple, and onion with bound

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service