Skip to Content
Merck
All Photos(3)

Key Documents

S1304

Sigma-Aldrich

Sodium acetate trihydrate

meets USP testing specifications

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

Assay

99.0-101.0% dry basis (HClO4 , titration)

form

crystalline

autoignition temp.

1112 °F

pH

7.5-9.2

solubility

water: soluble 408 g/L at 20 °C

application(s)

pharmaceutical (small molecule)

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Unraveling the effects of drying techniques on chaya leaves: Metabolomics analysis of nonvolatile and volatile metabolites, umami taste, and antioxidant capacity.: This study explores the impact of different drying methods on the biochemical composition of chaya leaves using sodium acetate trihydrate as a buffer component to measure antioxidant capacity. The research provides insights into optimizing processing methods to retain beneficial compounds and enhance flavor (Hutasingh N et al., 2024).

  • The role of urea in formation of the sodium acetate trihydrate (SAT)-urea eutectic liquid: a neutron diffraction and isotopic substitution study.: This paper discusses the use of sodium acetate trihydrate in creating a eutectic mixture with urea, which is characterized by neutron diffraction. The findings highlight the chemical′s utility in studying fluid properties and interactions at the molecular level, useful for designing novel solvents and materials (Byrne EL et al., 2024).

  • Fabrication of multi-functional gelatin/deep eutectic solvent/polyacrylonitrile nanofiber membranes via electrospinning.: This research uses sodium acetate trihydrate in the synthesis of advanced nanofiber structures for potential applications in filtration and biotechnology. The study showcases the adaptability of sodium acetate trihydrate in facilitating novel manufacturing processes (Cui Y et al., 2023).

  • Preparation, thermal storage properties and application of sodium acetate trihydrate/expanded graphite composite phase change materials.: Highlights the use of sodium acetate trihydrate in developing composite materials for energy storage. This study demonstrates the potential of these composites in enhancing the efficiency of thermal energy systems, which is crucial for developing sustainable energy solutions (Wang KW et al., 2023).

  • Improvement and enhancement of oligosaccharide production from Lactobacillus acidophilus using statistical experimental designs and its inhibitory effect on colon cancer.: This paper investigates the role of sodium acetate trihydrate in optimizing the fermentation conditions for the production of bioactive oligosaccharides. The study also examines the anti-cancer properties of these oligosaccharides, providing valuable data for future cancer research and probiotic development (Abo-Zaid GA et al., 2023).

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kit Frederiksen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 91, 9-15 (2015-01-18)
Polymeric film-forming systems (FFSs) are potential drug delivery systems for topical application to the skin. The FFSs form thin and transparent polymeric films in situ upon solvent evaporation. Their application convenience and cosmetic attributes, superior to conventional semi-solids, may offer
Claire E Mason et al.
Journal of the American Society for Mass Spectrometry, 24(2), 274-285 (2013-01-04)
Single-stranded DNA-binding proteins (SSBs) are ubiquitous oligomeric proteins that bind with very high affinity to single-stranded DNA and have a variety of essential roles in DNA metabolism. Nanoelectrospray ionization mass spectrometry (nanoESI-MS) was used to monitor subunit exchange in full-length
Horng-Ruey Chua et al.
Journal of critical care, 27(2), 138-145 (2012-03-24)
The purpose of the study was to determine the effects of Plasma-Lyte 148 (PL) vs 0.9% saline (NS) fluid resuscitation in diabetic ketoacidosis (DKA). A multicenter retrospective analysis of adults admitted for DKA to the intensive care unit, who received
Rishiram Ramanan et al.
FEBS letters, 587(4), 370-377 (2013-01-15)
Phenotypic and genotypic changes in Chlamydomonas reinhardtii BafJ5, a starchless mutant, with respect to lipid metabolism was studied in different trophic states under nitrogen (N) sufficient and limited conditions. Interestingly, cellular lipid content increased linearly with input acetate concentration with
Andrew D Shaw et al.
Annals of surgery, 255(5), 821-829 (2012-04-04)
To assess the association of 0.9% saline use versus a calcium-free physiologically balanced crystalloid solution with major morbidity and clinical resource use after abdominal surgery. 0.9% saline, which results in a hyperchloremic acidosis after infusion, is frequently used to replace

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service