RDD034
Urea
free-flowing, Redi-Dri™, Bioreagent, ≥98%
Synonym(s):
Carbamide, Carbonyldiamide
About This Item
Recommended Products
grade
reagent
Quality Level
product line
Redi-Dri™
Assay
≥98%
quality
Bioreagent
dilution
(for analytical testing)
mp
132-135 °C (lit.)
density
1.335 g/mL at 25 °C (lit.)
foreign activity
DNAse, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
Protease, none detected
RNAse, none detected
SMILES string
NC(N)=O
InChI
1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)
InChI key
XSQUKJJJFZCRTK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
- Redi-Dri™ packaging controls moisture for hygroscopic chemicals that commonly clump
- Anhydrous powder remains free-flowing and easy to use
- Safer to handle and weigh out for routine use in lab
- Eliminates clumps, maintaining product moisture, quality, and attributes
Other Notes
Legal Information
comparable product
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.
Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.
Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.
Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service