Skip to Content
Merck
All Photos(1)

Key Documents

Q0132

Quinine sulfate

meets USP testing specifications, monograph mol wt. 782.94 ((C20H24N2O2)2 . H2SO4 . 2H2O)

Synonym(s):

Quinine hemisulfate salt monohydrate, Quinine sulfate (2:1) (salt) dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2O2 · 0.5H2O4S · H2O
CAS Number:
Molecular Weight:
391.47
Beilstein:
6113937
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

mol wt

monograph mol wt. 782.94

mp

~225 °C (dec.) (lit.)

antibiotic activity spectrum

parasites

Mode of action

enzyme | inhibits

SMILES string

O.O.OS(O)(=O)=O.COc1ccc2nccc([C@@H](O)C3CC4CCN3C[C@@H]4C=C)c2c1.COc5ccc6nccc([C@@H](O)C7CC8CCN7C[C@@H]8C=C)c6c5

InChI

1S/2C20H24N2O2.H2O4S.2H2O/c2*1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;1-5(2,3)4;;/h2*3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;(H2,1,2,3,4);2*1H2/t2*13-,14-,19-,20+;;;/m00.../s1

InChI key

ZHNFLHYOFXQIOW-LPYZJUEESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Use to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites, the toxicity of heme (FP)-complexes and to inhibit mitochondrial ATP-regulated potassium channel.

Biochem/physiol Actions

Potassium channel blocker. Antimalarial, anticholinergic, antihypertensive, and hypoglycemic agent; alkaloid originally isolated from the Cinchona family of South American trees. Inhibits mitochondrial ATP-regulated potassium channel. Used to study the metabolism of biocrystalized heme, hemozoin, in malarial parasites and to study the toxicity of heme (FP)-complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Mooney et al.
Mucosal immunology, 7(6), 1302-1311 (2014-03-29)
Coinfection can markedly alter the response to a pathogen, thereby changing its clinical presentation. For example, non-typhoidal Salmonella (NTS) serotypes are associated with gastroenteritis in immunocompetent individuals. In contrast, individuals with severe pediatric malaria can develop bacteremic infections with NTS
Joe Briscoe et al.
Angewandte Chemie (International ed. in English), 54(15), 4463-4468 (2015-02-24)
New hybrid materials consisting of ZnO nanorods sensitized with three different biomass-derived carbon quantum dots (CQDs) were synthesized, characterized, and used for the first time to build solid-state nanostructured solar cells. The performance of the devices was dependent on the
Eva Woltmann et al.
Analytical and bioanalytical chemistry, 406(25), 6347-6362 (2014-08-12)
High production output of solid pharmaceutical formulations requires fast methods to ensure their quality. Likewise, fast analytical procedures are required in forensic sciences, for example at customs, to substantiate an initial suspicion. We here present the design and the optimization
Jie Ma et al.
Talanta, 143, 205-211 (2015-06-17)
Hollow cross-linked fluorescent carbon nitride nanoparticles (CNNPs) were fabricated via a facile one-pot solvothermal process. The obtained CNNPs were characterized by multiple analytical techniques including transmission electron microscopy (TEM), X-ray diffraction (XRD), X-ray photoelectron spectroscopy (XPS), solid-state nuclear magnetic resonance
Miaomiao Zhang et al.
International journal of nanomedicine, 10, 6943-6953 (2015-11-27)
X-ray computed tomography (CT) is the most commonly used imaging technique for noninvasive diagnosis of disease. In order to improve tissue specificity and prevent adverse effects, we report the design and synthesis of iodine-doped carbon dots (I-doped CDs) as efficient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service