Skip to Content
Merck
All Photos(1)

Key Documents

PHR1615

Supelco

Ritodrine Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ritodrine hydrochloride, 1-(4-Hydroxyphenyl)-2-[2-(4-hydroxyphenyl)ethylamino]propanol, N-(p-Hydroxyphenethyl)-4-hydroxynorephedrine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO3 · HCl
CAS Number:
Molecular Weight:
323.81
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. 586
traceable to USP 1604701

API family

ritodrine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.C[C@H](NCCc1ccc(O)cc1)[C@H](O)c2ccc(O)cc2

InChI

1S/C17H21NO3.ClH/c1-12(17(21)14-4-8-16(20)9-5-14)18-11-10-13-2-6-15(19)7-3-13;/h2-9,12,17-21H,10-11H2,1H3;1H/t12-,17-;/m0./s1

InChI key

IDLSITKDRVDKRV-XHXSRVRCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ritodrine Hydrochloride is a potent tocolytic drug and a selective γ2-adrenoceptor agonist, widely used for the control of premature labor.

Application

Ritodrine Hydrochloride may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3901 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel method for the spectrophotometric determination of isoniazid and ritodrine hydrochloride
Nagaraja P, et al.
Turkish Journal of Chemistry, 26(5), 743-750 (2002)
Spectrophotometric method for the determination of ritodrine hydrochloride and amoxicillin
Revanadiddappa DH, et al.
Analytical Sciences, 15(7), 661-664 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service