Skip to Content
Merck
All Photos(1)

Key Documents

PHR1584

Supelco

Donepezil Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Donepezil hydrochloride, (±)-2-[(1-Benzyl-4-piperidyl)methyl]-5,6-dimethoxy-1-indanone hydrochloride, 2,3-Dihydro-5,6-dimethoxy-2-{[1-(phenylmethyl)-4-piperidinyl]methyl}-1H-inden-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29NO3 · HCl
CAS Number:
Molecular Weight:
415.95
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1224981

API family

donepezil

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Cl-].N2(CCC(CC2)CC3Cc4c(cc(c(c4)OC)OC)C3=O)Cc1ccccc1.[H+]

InChI

1S/C24H29NO3.ClH/c1-27-22-14-19-13-20(24(26)21(19)15-23(22)28-2)12-17-8-10-25(11-9-17)16-18-6-4-3-5-7-18;/h3-7,14-15,17,20H,8-13,16H2,1-2H3;1H

InChI key

XWAIAVWHZJNZQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Donepezil Hydrochloride is known as a selective, reversible inhibitor of acetylcholinesterase enzyme. It exhibits therapeutic effect by raising the acetylcholine concentrations and improving the cholinergic function. It can be used in the treatment of mild to moderate dementia of the Alzheimer′s type.[1][2]

Application

Donepezil Hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations[1][3] and plasma samples[2][3] by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3756 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective HPLC assay of donepezil enantiomers with UV detection and its application to pharmacokinetics in rats.
Radwan MA, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 830(1), 114-119 (2006)
Quantitative determination of donepezil in human plasma by liquid chromatography/tandem mass spectrometry employing an automated liquid?liquid extraction based on 96-well format plates: Application to a bioequivalence study.
Apostolou C, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 848(2), 239-244 (2007)
A new HPLC method to determine Donepezil hydrochloride in tablets.
Pappa H, et al.
Journal of Pharmaceutical and Biomedical Analysis, 27(1-2), 177-182 (2002)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service